Skip to Content
Merck
All Photos(1)

Documents

ERMAC059

Aflatoxin G1 solution

3.78 μg/g in acetonitrile, ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O7
CAS Number:
Molecular Weight:
328.27
Beilstein:
1299768
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

concentration

3.78 μg/g in acetonitrile

application(s)

general analytical

format

single component solution

storage temp.

2-8°C

SMILES string

COc1cc2OC3OC=CC3c2c4OC(=O)C5=C(CCOC5=O)c14

InChI

1S/C17H12O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h3,5-6,8,17H,2,4H2,1H3

InChI key

XWIYFDMXXLINPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMAC059

Legal Information

ERM is a registered trademark of European Commission

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda M Aly et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(11), 3031-3034 (2010-05-18)
Different local and exported white cheese samples were collected from different markets in Jeddah during September 2008. Trace and heavy metals including Pb, Zn, Mn, Cu, Fe and Cd were analyzed using atomic absorption spectrometry. The concentration of the tested
Hongmei Zeng et al.
Applied microbiology and biotechnology, 90(2), 635-650 (2010-12-15)
In aflatoxin biosynthesis, aflatoxins G(1) (AFG(1)) and B(1) (AFB(1)) are independently produced from a common precursor, O-methylsterigmatocystin (OMST). Recently, 11-hydroxy-O-methylsterigmatocystin (HOMST) was suggested to be a later precursor involved in the conversion of OMST to AFB(1), and conversion of HOMST
Massimiliano Cuccioloni et al.
Analytical chemistry, 80(23), 9250-9256 (2009-06-25)
Aflatoxins are extremely toxic metabolites from Aspergillus species that can adulterate a wide range of human foodstuff. Herein, we propose a novel assay designed as an analytical test for aflatoxin B1 and G1 (AFB1 and AFG1, respectively) that could represent
Haitao Shen et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(9), 3222-3228 (2012-06-20)
Our previous studies showed intragastric administration of Aflatoxin G(1) (AFG(1)) could induce lung adenocarcinoma, which derived from alveolar type II cells (AT-II cells). AT-II cells contribute to Aflatoxin B(1) (AFB(1)) metabolism and also serve as the potential progenitor cells for
Zengning Li et al.
Toxicology letters, 195(2-3), 169-173 (2010-04-13)
Aflatoxins (AF) are the mycotoxin with the carcinogenic effect produced by the fungi like Aspergillus flavus, including AFB(1), B(2), G(1), G(2), M(1), and so on. The detection rate of Aflatoxin G(1) (AFG(1)) is very high in the diet of residents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service