Skip to Content
Merck
All Photos(1)

Key Documents

E1800000

Ethacrynic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2,3-Dichloro-4-[2-methylenebutyryl]phenoxy)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12Cl2O4
CAS Number:
Molecular Weight:
303.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ethacrynic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C(C(C(CC)=C)=O)=CC=C1OCC(O)=O

InChI

1S/C13H12Cl2O4/c1-3-7(2)13(18)8-4-5-9(12(15)11(8)14)19-6-10(16)17/h4-5H,2-3,6H2,1H3,(H,16,17)

InChI key

AVOLMBLBETYQHX-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethacrynic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Action and mode of action of a new original diuretic: ethacrynic acid].
M Salvador et al.
Journal de medecine de Bordeaux et du Sud-Ouest, 143(8), 1105-1118 (1966-08-01)
Toxicity of ethacrynic acid and furosemide.
L B Cooperman et al.
American heart journal, 85(6), 831-834 (1973-06-01)
Krzysztof Lukawski et al.
Pharmacological reports : PR, 66(6), 1100-1105 (2014-12-03)
Hypertension and heart failure belong to common comorbid conditions with epilepsy so drug interactions between antiepileptics and cardiovascular drugs are possible in clinical practice. The aim of this study was to evaluate the effects of angiotensin AT1 receptor antagonists (losartan
[Clinical pharmacology of diuretics with special reference to furosemide and ethacrynic acid].
J Prandota
Postepy higieny i medycyny doswiadczalnej, 34(1), 47-74 (1980-01-01)
Ethacrynic acid and related diuretics: relationship of structure to beneficial and detrimental actions.
D A Koechel
Annual review of pharmacology and toxicology, 21, 265-293 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service