Skip to Content
Merck
All Photos(2)

Key Documents

C7254

Sigma-Aldrich

Sodium citrate tribasic dihydrate

≥98%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98%

form

powder

pH

7.5-9 (25 °C, 29.4 g/L)

mp

>300 °C (lit.)

solubility

water: soluble 100 mg/mL

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Deciphering the orthorhombic crystal structure of a novel NEIL1 nanobody with pseudo-merohedral twinning.: Sodium citrate tribasic dihydrate was utilized as a buffering agent in the crystallization process, facilitating the analysis of a NEIL1 nanobody′s crystal structure, which has implications for understanding DNA repair mechanisms (Thompson et al., 2023).

  • The coiled-coil domain of glycosomal membrane-associated Leishmania donovani PEX14: cloning, overexpression, purification and preliminary crystallographic analysis.: Sodium citrate tribasic dihydrate played a critical role in the purification and crystallization of the coiled-coil domain of PEX14, advancing research on the molecular mechanisms of Leishmania donovani (Shakya and Pratap, 2020).

  • Determination of Mycotoxins in Brown Rice Using QuEChERS Sample Preparation and UHPLC-MS-MS.: This study employs sodium citrate tribasic dihydrate in the QuEChERS method for efficient extraction of mycotoxins, enhancing the accuracy of mycotoxin determination in brown rice samples (Jettanajit and Nhujak, 2016).

  • Electroless Cu Plating on Anodized Al Substrate for High Power LED.: Sodium citrate tribasic dihydrate was used as a complexing agent in the electroless copper plating process, which improved the performance and reliability of high power LEDs (Rha and Lee, 2015).

  • Crystallization and preliminary X-ray analysis of a novel type of lipolytic hydrolase from Bacillus licheniformis.: Sodium citrate tribasic dihydrate was essential in the crystallization of a novel lipolytic hydrolase, aiding in the structural analysis and potential industrial applications of this enzyme (Ju et al., 2014).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Savchuk et al.
Steroids, 149, 108417-108417 (2019-06-01)
Steroidogenic potential of the human fetal kidney (hFK) at the end of first trimester is poorly investigated. Little is known about the ontogeny of steroidogenic enzymes and activities of steroidogenic pathways in the hFK at early pregnancy. Our aim was
Ruonan Xu et al.
Cancer cell international, 18, 138-138 (2018-09-18)
Both multiple myeloma (MM) and systemic lupus erythematosus (SLE) are associated with abnormal production of plasma cells, although their pathological mechanism of each disease is different. The main characteristic of both diseases is uncontrolled differentiation of B cells into plasmablast/plasma
Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent
Ruben de Vries et al.
Journal of dairy science, 98(12), 8678-8687 (2015-10-12)
The aim of this study was to evaluate the effect of shortening or omitting the dry period of dairy cows on milk casein composition. For this study, we analyzed milk samples of 90 cows with a dry period of 0
Jun Zhao et al.
Nanotechnology, 25(40), 405101-405101 (2014-09-12)
The treatment of liver injuries or hepatocellular carcinoma (HCC) has been hindered by the lack of efficient drug delivery. Even with the help of nanoparticles or other synthetic delivering agents, a large portion of the dose is still sequestered in

Articles

Citrate is one of many cell culture components that affects the performance of serum-free, protein-free and animal protein-free cell culture systems used for biomanufacturing heterologous proteins such as monoclonal antibodies.

Fast assessment of antibody-dependent cell-mediated cytotoxicity (ADCC) and glycoform pattern of a therapeutic antibody rituximab by FcγRIIIa affinity chromatography.

Fast assessment of antibody-dependent cell-mediated cytotoxicity (ADCC) and glycoform pattern of a therapeutic antibody rituximab by FcγRIIIa affinity chromatography.

Fast assessment of antibody-dependent cell-mediated cytotoxicity (ADCC) and glycoform pattern of a therapeutic antibody rituximab by FcγRIIIa affinity chromatography.

See All

Protocols

Enzymatic Assay of Acid Phosphatase (EC 3.1.3.2)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service