Skip to Content
Merck
All Photos(1)

Key Documents

71405

Sigma-Aldrich

Sodium citrate tribasic dihydrate

puriss. p.a., ACS reagent, ≥99.0% (NT)

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
form:
powder
solubility:
water: soluble 29.4 g/L at 20 °C

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.0% (NT)

form

powder

impurities

11-13% water (KFT)

pH

7.0-9.0 (25 °C, 50 mg/mL in H2O)

mp

>300 °C (lit.)

solubility

water: soluble 29.4 g/L at 20 °C

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤10 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Sodium citrate tribasic dihydrate is the tribasic dihydrate sodium salt of citric acid.

Application

  • Kinetic Study of Silver Nanoparticle Formation: This study examines the influence of varying concentrations of sodium citrate tribasic dihydrate in the synthesis of silver nanoparticles. The sodium citrate acts as a reducing agent, impacting the formation and stability of the nanoparticles. The detailed kinetic analysis contributes to understanding the nucleation and growth mechanisms of silver nanoparticles in the presence of sodium citrate tribasic dihydrate (SA Diaz - The Undergraduate Research Journal at the University of Northern Colorado, 2019).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrocatalytic activity of carbon-supported Pt-Au nanoparticles for methanol electro-oxidation.
Park IS, et al.
Electrochimica Acta, 52(18), 5599-5605 (2007)
Sukdeb Pal et al.
Applied and environmental microbiology, 73(6), 1712-1720 (2007-01-31)
In this work we investigated the antibacterial properties of differently shaped silver nanoparticles against the gram-negative bacterium Escherichia coli, both in liquid systems and on agar plates. Energy-filtering transmission electron microscopy images revealed considerable changes in the cell membranes upon
Toxicity of silver nanoparticles?nanoparticle or silver ion?
Beer C, et al.
Toxicology Letters, 208(3), 286-292 (2012)
Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent
Ruben de Vries et al.
Journal of dairy science, 98(12), 8678-8687 (2015-10-12)
The aim of this study was to evaluate the effect of shortening or omitting the dry period of dairy cows on milk casein composition. For this study, we analyzed milk samples of 90 cows with a dry period of 0

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service