Skip to Content
Merck
All Photos(1)

Key Documents

A3669

Supelco

Acebutolol hydrochloride

analytical standard

Synonym(s):

N-(3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl)butanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O4 · HCl
CAS Number:
Molecular Weight:
372.89
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCCC(=O)Nc1ccc(OCC(O)CNC(C)C)c(c1)C(C)=O

InChI

1S/C18H28N2O4.ClH/c1-5-6-18(23)20-14-7-8-17(16(9-14)13(4)21)24-11-15(22)10-19-12(2)3;/h7-9,12,15,19,22H,5-6,10-11H2,1-4H3,(H,20,23);1H

InChI key

KTUFKADDDORSSI-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

Acebutolol hydrochloride is a β-adrenergic receptor blocker. It is used in the treatment of hypertension, cardiac arrhythmias and angina pectoris.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2013 Nurse's Drug Handbook (2012)
Donatella Ottaviani et al.
Journal of medical microbiology, 62(Pt 1), 161-164 (2012-09-25)
We report a severe case of travellers' diarrhoea in a patient returning from Ecuador to Italy with the concomitant presence of Aeromonas veronii biovar sobria and Vibrio parahaemolyticus in their faeces. Based on diagnostic results, epidemiological information and the clinical
Iu M Alekseeva et al.
Kardiologiia, 47(12), 24-34 (2008-02-12)
Beta-blocking agents (BB) decrease mortality particularly sudden in post-myocardial infarction patients and in patients with chronic heart failure that is closely related to heart rate (HR) lowering at rest. The hypothesis exists that BB decrease mortality due to increase of
Stéphane Rinfret et al.
American heart journal, 153(2), 224-230 (2007-01-24)
Long-term treatment with beta-blockers reduces mortality after acute myocardial infarction (AMI). Whether beta-blockers exert a class effect is unknown. We analyzed mortality after AMI in Canadian patients 65 years or older who were discharged from hospital with a diagnosis of
Stevan Armaković et al.
Journal of molecular modeling, 18(9), 4491-4501 (2012-05-31)
The aim of this study is to investigate the active components of representative drugs for blood pressure regulation by applying quantum mechanical computer codes and comparison of the same for the sake of obtaining knowledge about the properties associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service