Skip to Content
Merck
All Photos(1)

Key Documents

94000

Supelco

Undecane

analytical standard

Synonym(s):

n-Undecane, Hendecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
Beilstein:
1697099
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.4 (vs air)

vapor pressure

<0.4 mmHg ( 20 °C)

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

cycloparaffins
isoparaffin

refractive index

n20/D 1.417 (lit.)
n20/D 1.418

bp

196 °C (lit.)

mp

−26 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCC

InChI

1S/C11H24/c1-3-5-7-9-11-10-8-6-4-2/h3-11H2,1-2H3

InChI key

RSJKGSCJYJTIGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Undecane is obtained from the refining of petroleum. It is a component of gasoline and is generally used in organic synthesis, petroleum research and as a distillation chaser.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Undecane has been used as a standard in quantitatively determining the presence of iso-furans and F2-isoprostanes in biological fluids and tissues, using gas chromatography coupled with mass spectrometry (GC/MS).
Undecane has been used as a standard in quantitatively determining the presence of iso-furans and F2-isoprostanes in biological fluids and tissues, using gas chromatography coupled with mass spectrometry(GC/MS).

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Measurement of F 2-isoprostanes and isofurans using gas chromatography?mass spectrometry
Milne.LG, et al.
Free Radical Biology & Medicine, 59, 36-44 (2013)
Patty's Toxicology, 6(1) (2012)
Subodh P Jagtap et al.
Journal of the American Chemical Society, 134(16), 7176-7185 (2012-03-30)
In this work, a bicyclo[4.4.1]undecane scaffold is used to hold oligo(phenylene ethynylene) units in a cofacially stacked arrangement along the entire length of the conjugated units. We study the impact that the resulting strong interchain interactions have on the photophysical
Hema Tresa Varghese et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(5), 513-522 (2010-05-18)
Fourier-transform (FT)-Raman and FT-infrared spectrum of 4-(3-bromopropyl)-4-azatricyclo [5.2.2.0(2,6)]undecane-3,5,8-trione were recorded and analyzed. The vibrational wavenumbers were examined theoretically using the Gaussian03 set of quantum chemistry codes, and the normal modes were assigned by potential energy distribution (PED) calculations. The first
C Yohannan Panicker et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(5), 1559-1565 (2010-03-17)
Fourier-transform-Raman and infrared spectrum of 4-azatricyclo [5.2.2.0(2,6)] undecane-3,5,8-trione were recorded and analyzed. The vibrational wavenumbers were examined theoretically using the Gaussian03 set of quantum chemistry codes, and the normal modes were assigned by potential energy distribution (PED) calculations. The first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service