Skip to Content
Merck
All Photos(1)

Key Documents

91490

Supelco

Tridecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CH3
CAS Number:
Molecular Weight:
184.36
Beilstein:
1733089
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

6.4 (vs air)

vapor pressure

1 mmHg ( 59.4 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.425 (lit.)
n20/D 1.426

bp

110-112 °C/12 mmHg (lit.)
234 °C (lit.)

mp

−6-−4 °C (lit.)

density

0.756 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCC

InChI

1S/C13H28/c1-3-5-7-9-11-13-12-10-8-6-4-2/h3-13H2,1-2H3

InChI key

IIYFAKIEWZDVMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tridecane may be used as internal standard in the analysis of isomeric ratio of diethyl[(1E)-hexa-1,5-dienyl]methylsilane and diethyl[(1E,4E)-hexa-1,4-dienyl]methylsilane obtained via dehydrogenative silylation of hexa-1,5-diene using gas liquid chromatography(GLC).[1][2]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F

Flash Point(C)

79 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ash M et al.
Handbook of Green Chemicals (2004)
Adolfsson H, et al. et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 4 (2014)
Bo Li et al.
Chemical communications (Cambridge, England), 49(12), 1187-1189 (2013-01-04)
A phytochemical investigation of the cone of Pseudolarix amabilis led to the isolation of pseudolarenone (), a structurally novel pentacyclic (5/11/5/6/5) nortriterpenoid lactone with an unprecedented carbon skeleton featuring a unique bicyclo[8.2.1]tridecane core. Its structure and absolute configurations were elucidated
Dorit Eliyahu et al.
Journal of chemical ecology, 38(11), 1358-1365 (2012-10-20)
One characteristic of true bugs (Heteroptera) is the presence of dorsal abdominal glands in the immature nymphal stages. These glands usually produce defensive chemicals (allomones) that vary among taxa but are still similar in closely related groups. Knowledge of the
Jörg P Hehn et al.
Organic letters, 13(7), 1892-1895 (2011-03-09)
Starting from tetronate 1 (R = CH(2)OH), a short photochemical access to the 3,12-dioxatricyclo[8.2.1.0(6,13)]tridecane-skeleton 2 of briarellin and asbestinin diterpenes has been explored. In the course of these studies, a number of surprising observations were made. For example, a two-step

Protocols

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service