Skip to Content
Merck
All Photos(2)

Key Documents

89063

Supelco

4-Bromo-α-cyanocinnamic acid

matrix substance for MALDI-MS, ≥95.0% (HPLC)

Synonym(s):

3-(4-Bromophenyl)-2-cyano-2-propenoic acid, p-Bromo-α-cyanocinnamic acid, p-Bromobenzylidenecyanoacetic acid, BrCCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6BrNO2
CAS Number:
Molecular Weight:
252.06
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥95.0% (HPLC)

form

powder

analyte functional class(es)

drugs of abuse, ionic liquids (quantification)

analyte chemical class(es)

chlorinated lipids, lipids, peptides, phospholipids, phosphopeptides

technique(s)

MALDI-MS: suitable

solubility

methanol: 100 mg/10 mL, clear, colorless to light yellow

SMILES string

OC(/C(C#N)=C/C1=CC=C(Br)C=C1)=O

InChI

1S/C10H6BrNO2/c11-9-3-1-7(2-4-9)5-8(6-12)10(13)14/h1-5H,(H,13,14)/b8-5+

InChI key

REWONLOPJHWIMK-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • suitable as MALDI-Matrix material
  • suitable for collision-induced dissociation MS/MS (CID-MS/MS)

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimitrios G Papasotiriou et al.
Journal of proteome research, 9(5), 2619-2629 (2010-04-07)
Peptide Mass Fingerprinting (PMF) of tryptically in-gel digested samples is a well-established protein identification technique for MALDI mass spectrometry but an in-depth PMF evaluation for in-gel digestions of less specific enzymes is still missing. This study demonstrates that the MALDI-LTQ-Orbitrap
Thorsten Jaskolla et al.
Journal of the American Society for Mass Spectrometry, 20(5), 867-874 (2009-02-10)
Phosphatidylethanolamines (PEs) are abundant lipid constituents of the cellular membrane. The amino group of PEs exhibits high reactivity with hypochlorous acid that is generated under inflammatory conditions in vivo. The analysis of the resulting PE mono- and dichloramines is of
Marcel Wiegelmann et al.
Analytical and bioanalytical chemistry, 405(22), 6925-6932 (2012-10-16)
A high analytical sensitivity in ultraviolet matrix-assisted laser desorption ionization mass spectrometry (MALDI-MS) is only achieved if the laser wavelength corresponds to a high optical absorption of the matrix. Laser fluence and the physicochemical properties of the compounds, e.g., the
Thorsten W Jaskolla et al.
Journal of proteome research, 8(7), 3588-3597 (2009-05-14)
The performance of the recently developed 4-chloro-alpha-cyanocinnamic acid (Cl-CCA) matrix-assisted laser desorption ionization mass spectrometry (MALDI MS) matrix was investigated in comparison to the most widely used matrix alpha-cyano-4-hydroxycinnamic acid (CHCA). For this purpose, in-solution digestions of standard proteins in
Jens Soltwisch et al.
Journal of the American Society for Mass Spectrometry, 24(10), 1477-1488 (2013-08-15)
The success of matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) as a widely employed analytical tool in the biomolecular sciences builds strongly on an effective laser-material interaction that is resulting in a soft co-desorption and ionization of matrix and imbedded biomolecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service