Skip to Content
Merck

Skip To

A8471

ProteoMass Albumin MALDI-MS Standard

vial of 10 nmol, average mol wt 66,429.09 Da by calculation

Synonym(s):

Albumin, bovine, mass spec standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
EC Number:
232-936-2
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

average mol wt 66,429.09 Da by calculation

Quality Segment

packaging

vial of 10 nmol

format

neat

Application

ProteoMass Albumin MALDI-MS Standard has been used as a standard in matrix-assisted laser desorption/ionization-mass spectrometry (MALDI-MS)[1] and mass spectrometry.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8971A8846B4181
format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

packaging

vial of 10 nmol

packaging

vial of 10 nmol

packaging

vial of 10 nmol

packaging

vial of 10 nmol

mol wt

average mol wt 66,429.09 Da by calculation

mol wt

(M+H+) 16,952.27 Da by calculation

mol wt

monoisotopic mol wt 1,045.5423 Da

mol wt

monoisotopic mol wt 756.3997 Da


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters