Skip to Content
Merck
All Photos(3)

Key Documents

861111

Sigma-Aldrich

Bismarck Brown Y

certified by the Biological Stain Commission, certified by the BSC, Dye content 50 %

Synonym(s):

4,4′-(m-Phenylenebisazo)bis-m-phenylenediamine dihydrochloride, Basic Brown 1, Bismarck Brown B, Vesuvine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N8 · 2HCl
CAS Number:
Molecular Weight:
419.31
Colour Index Number:
21000
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

Agency

certified by the BSC

form

powder

composition

Dye content, 50%

technique(s)

microbe id | staining: suitable

solubility

H2O: 10 mg/mL, clear to turbid, red-orange to red

λmax

457 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.Cl.Nc1ccc(\N=N\c2cccc(c2)\N=N\c3ccc(N)cc3N)c(N)c1

InChI

1S/C18H18N8.2ClH/c19-11-4-6-17(15(21)8-11)25-23-13-2-1-3-14(10-13)24-26-18-7-5-12(20)9-16(18)22;;/h1-10H,19-22H2;2*1H/b25-23+,26-24+;;

InChI key

MCZVRBLCRZWFJH-SPBSJSFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bismarck Brown Y is used for the staining of plant samples, including stem, root, starch granules and epidermis.

Suitability

Certified for use in staining goblet cells and cartilage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Laboratory and Clinical Medicine, 26, 1200-1200 (1941)
R.D. Lillie Ed.
Conn?s Biological Stains, 145-145 (1977)
The use of Bismarck brown Y in some new stain schedules.
Blaydes GW
Stain Technology, 14.3, 105-110 (1939)
E T Graham et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 73(4), 178-185 (1998-09-15)
A staining solution containing alcian blue 8GX, Bismarck brown Y and safranin O was prepared with 0.1 M sodium acetate buffer, pH 5.0. Paraffin was extracted with MicroClear solvent from 10 microm tissue sections mounted on slides. Paraffin solvent was
B G Prakash Kumar et al.
Journal of hazardous materials, 136(3), 922-929 (2006-03-03)
Activated carbon was produced from a biowaste product, rubberwood sawdust (RWSD) using steam in a high temperature fluidized bed reactor. Experiments were carried out to investigate the influence of various process parameters such as activation time, activation temperature, particle size

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service