Skip to Content
Merck
All Photos(3)

Key Documents

861286

Sigma-Aldrich

Orange G

certified by the Biological Stain Commission, Dye content 80 %

Synonym(s):

1-Phenylazo-2-naphthol-6,8-disulfonic acid disodium salt, 7-Hydroxy-8-phenylazo-1,3-naphthalenedisulfonic acid disodium salt, Acid Orange 10, Wool Orange 2G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2Na2O7S2
CAS Number:
Molecular Weight:
452.37
Colour Index Number:
16230
Beilstein:
4120705
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, 80%

technique(s)

microbe id | staining: suitable

color

orange to very dark orange, and Red to Very Dark Red and Orange-Red and Red-Orange and Dark Red-Orange

solubility

H2O: 1 mg/mL

λmax

475 nm

ε (extinction coefficient)

≥10000 at 329-331 nm in H2O
≥17000 at 473-479 nm in H2O
≥20500 at 246-250 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1ccc2cc(cc(c2c1\N=N\c3ccccc3)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H12N2O7S2.2Na/c19-13-7-6-10-8-12(26(20,21)22)9-14(27(23,24)25)15(10)16(13)18-17-11-4-2-1-3-5-11;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2/b18-17+;;

InChI key

HSXUHWZMNJHFRV-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Collagen stain for connective tissue. Used with Orcein, Alizarin Blue 2B, and Fast Green FCF in Kornhauser′s quadruple stain for most elementary structures of tissues.

Biochem/physiol Actions

Orange G is used in collagen stain for connective tissue. It is used with Orcein, Alizarin Blue 2B, and Fast Green FCF in Kornhauser′s quadruple stain for most elementary structures of tissues. It is also used along with acid fuchsin and light green in masson trichrome stain for the staining of cellular structures, red blood cells and muscle cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kierszenbaum AL and Tres L
Histology and Cell Biology: An Introduction to Pathology (2015)
Asli Yuksel et al.
Journal of hazardous materials, 190(1-3), 1058-1062 (2011-03-29)
Complete degradation of azo dye Orange G was studied using a 500 mL continuous flow reactor made of SUS 316 stainless steel. In this system, a titanium reactor wall acted as a cathode and a titanium plate-type electrode was used
M Arulkumar et al.
Journal of hazardous materials, 186(1), 827-834 (2011-01-15)
Thespesia populnea is a large tree found in the tropical regions and coastal forests of India. Its pods were used as a raw material for the preparation of activated carbon. The prepared activated carbon was used for the adsorptive removal
Arumugam Sivasamy et al.
Current microbiology, 62(2), 351-357 (2010-07-21)
Biosorption is an eco-friendly and cost-effective method for treating the dye house effluents. Aspergillus niger and Trichoderma sp. were cultivated in bulk and biomasses used as biosorbents for the biosorption of an azo dye Orange G. Batch biosorption studies were
G Thennarasu et al.
Environmental science and pollution research international, 19(7), 2755-2765 (2012-02-23)
The photocatalytic degradation of Orange G (OG) dye has been investigated using synthesised nanocrystalline ZnO as a photocatalyst and sunlight as the irradiation source. The formation of ZnO prepared from its precursor was confirmed through FT-IR and powder X-ray diffraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service