Skip to Content
Merck
All Photos(1)

Documents

83913

Sigma-Aldrich

RNA isolation kit

BioUltra, for molecular biology

Synonym(s):

Ribonucleic acid isolation kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

storage temp.

2-8°C

General description

RNA isolation kit utilizes a rapid single-step extraction (guanidine thiocyanate/phenol/chloroform extraction) to isolate undegraded total RNA; the kit contains enough reagents to isolate total RNA from at least 7g of tissue.
Content: Denaturating solution, phenol saturated with H2O, β-mercaptoethanol, chloroform:isoamyl alcohol, 2 M sodium acetate, isopropanol, and instruction manual.

Other Notes

Kit used for the isolation of RNA

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Oral - STOT RE 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Heart, Liver,Kidney, Nervous system,Kidney,Liver,Skin

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J McKenzie et al.
Journal of sports science & medicine, 10(2), 385-392 (2011-01-01)
Genes can be activated or inhibited by signals within the tissues in response to an acute bout of exercise. It is unclear how a particular aerobic exercise bout may influence two muscles with similar actions to the activity. Therefore, the
Muhammad Imran Khan et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-21)
Alzheimer's disease (AD) is the most frequent type of dementia affecting memory, thinking and behaviour. The major hallmark of the disease is pathological neurodegeneration due to abnormal aggregation of Amyloid beta (Aβ) peptides generated by β- and γ-secretases via amyloidogenic
Q Zeng et al.
Current molecular medicine, 17(7), 509-517 (2018-02-14)
The basic helix-loop-helix (bHLH) protein families are a large class of transcription factors, which are associated with cell proliferation, tissue differentiation, and other important development processes. We reported that the Nuclear localized protein-1 (Nulp1) might act as a novel bHLH
Kuan-Bing Chen et al.
Cell death & disease, 12(8), 748-748 (2021-07-30)
Chemotherapy remains the primary treatment of advanced solid cancer, including lung cancer. However, as first-line treatment, cisplatin-based therapy is restricted by the frequent development of drug resistance. Increasing data showed that the programmed cell death protein ligand 1 (PD-L1) plays
P Chomczynski et al.
Analytical biochemistry, 162(1), 156-159 (1987-04-01)
A new method of total RNA isolation by a single extraction with an acid guanidinium thiocyanate-phenol-chloroform mixture is described. The method provides a pure preparation of undegraded RNA in high yield and can be completed within 4 h. It is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service