Skip to Content
Merck

Skip To

RTN9604

GenElute 96 Well Total RNA Purification Kit

greener alternative

4 x 96 well plate, sufficient for 4 96-well plate purifications

Synonym(s):

High Throughput Purification, RNA Purification

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
41105501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 4 96-well plate purifications

quality

4 x 96 well plate

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

RNA purification: suitable

input

mammalian cells
mammalian tissue

test parameters

: ~70 min kit run time per plate, sample volume: up to 10^7 cells or 30 mg of tissue

greener alternative category

storage temp.

15-25°C

General description

GenElute 96 Well Total RNA Purification Kit allows for the high-throughput purification of total RNA from cells or tissue. The kit utilizes the advantages of silica membrane technology and eliminates the use of hazardous organic compounds such as phenol and chloroform.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

The purified total RNA is ready for use in any enzymatic downstream application.

Biochem/physiol Actions

Sigma′s GenElute 96 Well Total RNA Purification Kit provides a simple and convenient way to isolate total RNA from cells or tissue. The cells or tissue are lysed in a solution containing large amounts of chaotropic salt which immediately inactivates RNases that are present in almost all biological materials. The addition of a Wash Buffer provides the necessary binding conditions for the RNA to be adsorbed to the silica membrane. The contaminating DNA which may be bound to the silica membrane is removed by directly applying RNase-free DNase. The remaining salts, proteins, and other cellular debris are removed by additional washing steps. The final step is to elute the purified RNA in the RNAase-free water that is supplied in the kit.

Features and Benefits

  • Unique plate design offers low risk of cross-contamination
  • RNA can be used for any kind of downstream enzymatic reaction
  • Processing possible under with a vacuum or centrifuge
  • Suitable for manual and automated processing

Other Notes

Application notes:
Method of purification:Silica-membrane technology
Compatible with Manual or automated, vacuum or centrifugation
Efficient recovery of > 200 nt
Typical yield20 μg (2 x 106 HeLa cells),
20 μg (20 mg mouse liver)
Typical concentration50–200 ng/μL
Elution volume50–130 μL
Preparation time70 min/plate
Binding capacity100 μg
RNA ratio28S/18S approx. 2.1
A260/A2801.90–2.10
Typical RIN (RNA > 9 (cells)
integrity number)≥ 7 (tissue)

Sufficient for:
< 107 cultured cells (centrifugation)
< 30 mg tissue (centrifugation)
saliva

Related products:
Prod #Related prod #
RTN9604RTN9602
RTN9602RTN9604

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
GenElute is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PCR9604RTN10RTN70
technique(s)

RNA purification: suitable

technique(s)

DNA purification: suitable

technique(s)

RNA purification: suitable

technique(s)

RNA purification: suitable

usage

sufficient for 4 96-well plate purifications

usage

sufficient for 4 96-well plate purifications

usage

sufficient for 10 purifications

usage

sufficient for 70 purifications

input

mammalian cells
mammalian tissue

input

sample(s) (PCR)

input

mammalian cells
mammalian tissue

input

mammalian tissue
mammalian cells

sustainability

Greener Alternative Product

sustainability

-

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

greener alternative category

Aligned,

greener alternative category

-

greener alternative category

, Aligned

greener alternative category

Aligned,

test parameters

: ~70 min kit run time per plate, sample volume: up to 10^7 cells or 30 mg of tissue

test parameters

: <45 min kit run time, sample volume: 100 μL

test parameters

: <30 min kit run time, sample volume: up to 10^7 cells or 40 mg of tissue

test parameters

: <30 min kit run time, sample volume: up to 10^7 cells or 40 mg of tissue


Kit Components Only

Product No.
Description

  • RNA Wash Buffer 2, Concentrate

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Can the lysates, after the cells are lysed in the kits lysis buffer, be stored at -80°C, or is it necessary for the RNA purification to be carried out immediately?

    1 answer
    1. Following disruption using Product No. RTN9604 - GenElute™ 96 Well Total RNA Purification Kit, the samples can be stored at -70°C in Lysis Buffer. Frozen samples are stable for up to 6 months. Before commencing the RNA isolation, frozen samples in Lysis Buffer should be completely thawed and centrifuged.

      Helpful?

Reviews

No rating value

Active Filters