Skip to Content
Merck
All Photos(1)

Key Documents

79899

Supelco

Piperitone

analytical standard

Synonym(s):

3-Carvomenthenone, 6-Isopropyl-3-methyl-2-cyclohexen-1-one, p-Menth-1-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (mixture of enantiomers, predominantly (-)-enantiomer, GC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

CC(CCC1C(C)C)=CC1=O

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h6-7,9H,4-5H2,1-3H3

InChI key

YSTPAHQEHQSRJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Piperitone is a monocyclic monoterpenoid, which occurs in eucalyptus oil. It is a good source of menthone and thymol.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

182.3 °F - closed cup

Flash Point(C)

83.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organic Chemistry, Volume 2: Stereochemistry And The Chemistry Natural Products, 5/E (1956)
Essential Oil Safety - E-Book: A Guide for Health Care Professionals (2013)
Terpenoids (2009)
Magali Picard et al.
Journal of agricultural and food chemistry, 64(40), 7576-7584 (2016-10-01)
Piperitone was recently identified in red Bordeaux wines, and this study was designed to further explore its contribution to wine aroma. Firstly, a geographical origin effect was detected within the Bordeaux region (left versus right banks of the Gironde estuary)
Alexandre Pons et al.
Food chemistry, 206, 191-196 (2016-04-05)
The present study concerns the search for new aroma compounds associated with the flavor of aged and prematurely aged red wines. Sensory descriptive analysis associated with gas chromatography-olfactometry was first performed to find specific odoriferous zones. One of the zones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service