Skip to Content
Merck
All Photos(1)

Key Documents

422770

Sigma-Aldrich

Menthyl acetate

97%

Synonym(s):

(±)-Menthol acetate, (±)-Menthyl acetate, dl-Menthyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

228-229 °C (lit.)

density

0.922 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(C)C1CCC(C)CC1OC(C)=O

InChI

1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3

InChI key

XHXUANMFYXWVNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Menthyl acetate (MA), a monoterpene is one of the main constituent of the peppermint essential oil obtained from the leaves of Mentha piperita. It has been reported to show ambulation-promoting effect in mice. Its fumigant and repellent activity has been evaluated on first-instar nymphs of the bloodsucking bug Rhodnius prolixus. The decomposition of MA in an atmospheric flow reactor and a low-pressure flat flame has been investigated by molecular-beam mass spectrometry (MBMS).

Menthyl acetate is a monoterpene that is widely utilized as a flavoring agent in the food and beverage industry.

Application

Menthyl acetate may be used in the preparation of 3-amino-2-hydroxyalkanoates via lithium amide-supported Mannich-type reaction of its lithium enolate with PMP-arylaldimines (PMP=p-methoxyphenyl) followed by in situ oxidation with oxaziridine.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V Sfara et al.
Journal of medical entomology, 46(3), 511-515 (2009-06-06)
The aim of this study was to evaluate the fumigant and repellent activity of five essential oils (from eucalyptus, geranium, lavender, mint, and orange oil) and seven monoterpenes (eucalyptol, geraniol, limonene, linalool, menthone, linalyl acetate, and menthyl acetate) on first-instar
T Umezu et al.
Pharmacology, biochemistry, and behavior, 69(3-4), 383-390 (2001-08-18)
Various plant-derived essential oils (EOs) have traditionally been used in the treatment of mental disorders, despite a lack of scientific evidence. In a previous study, we demonstrated that certain EOs possess behavioral effects, a finding that supports our original hypotheses
Faqir Muhammad et al.
Cutaneous and ocular toxicology, 36(3), 237-252 (2016-11-09)
A large number of cosmetics and topical pharmaceuticals contain compounds of natural origin. There is a rising concern if these compounds can interact with the activity of other topically applied components in these formulations. The current study demonstrates modulation of
Marina D Soković et al.
Molecules (Basel, Switzerland), 14(1), 238-249 (2009-01-13)
The potential antifungal effects of Thymus vulgaris L., Thymus tosevii L., Mentha spicata L., and Mentha piperita L. (Labiatae) essential oils and their components against 17 micromycetal food poisoning, plant, animal and human pathogens are presented. The essential oils were
Kinetic studies of methyl acetate pyrolysis and oxidation in a flow reactor and a low-pressure flat flame using molecular-beam mass spectrometry.
Yang X, et al.
Proceedings of the Combustion Institute, 35(1), 491-498 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service