Skip to Content
Merck
All Photos(1)

Key Documents

78943

Supelco

Butylated hydroxyanisole

analytical standard

Synonym(s):

Butylated hydroxyanisole, E320, 2(3)-t-Butyl-4-hydroxyanisole, 2(3)-t-Butylhydroquinone monomethyl ether, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(OCH3)OH
CAS Number:
Molecular Weight:
180.24
EC Number:
MDL number:
UNSPSC Code:
85151701
E Number:
E320
NACRES:
NA.24

Quality Level

grade

analytical standard

vapor density

6.2 (vs air)

Assay

≥99.0% (sum of isomers, GC)

autoignition temp.

599 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

58-60 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

O(C)c1cc(c(cc1)O)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butylated hydroxyanisole (BHA), a lipophilic compound, is reported to exhibit antimicrobial activity against molds and Gram-positive bacteria. It also finds applications as a direct/indirect additive in adhesives, lubricants, food packaging materials, defoaming agents, as a preservative in cosmetic products, antioxidant in petroleum products including jet fuels, cutting oils, rubber latex gloves, paints, lacquers, adhesive hardeners, cleaning agents, thinners, plastics, etc. It is also widely used as an antioxidant in food industry.††
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Butylated hydroxyanisole may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Food samples using square-wave voltammetry (SWV), further confirmed by high-performance liquid chromatography technique (HPLC) and capillary electrophoresis (CE) coupled with end-column amperometric detection.
  • Cosmetics using fluorescence spectroscopy with second-order calibration, further confirmed by HPLC technique.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

241.9 °F - Pensky-Martens closed cup

Flash Point(C)

116.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicology and biochemistry of butylated hydroxyanisole and butylated hydroxytoluene
Branen.L.A
Journal of the American Oil Chemists' Society, 52, 59-63 (1975)
Quantitative determination of butylated hydroxyanisole and n-propyl gallate in cosmetics using three-dimensional fluorescence coupled with second-order calibration
Wang Y-J, et al.
Talanta, 116, 347-353 (2013)
CRC Handbook of Food, Drug, and Cosmetic Excipients (2018)
Safety assessment of butylated hydroxyanisole and butylated hydroxytoluene as antioxidant food additives
Williams.M.G, et al.
Food And Chemical Toxicology, 37, 1027-1038 (1999)
Capillary electrophoresis-amperometric determination of antioxidant propyl gallate and butylated hydroxyanisole in foods
Xiang Q, et al.
Analytical Sciences, 23(6), 713-717 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service