Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
2-[(CH3)3C]-4-(CH3O)C6H3OH
CAS Number:
Molecular Weight:
180.24
UNSPSC Code:
12352104
E Number:
E320
EC Index Number:
246-563-8
MDL number:
vapor pressure
1.33 hPa ( 105 °C)
Quality Segment
form
solid
pH
4.8 (25 °C, 10 g/L in H2O)
bp
263 °C/1013 hPa
mp
48-63 °C
transition temp
flash point 142.1 °C
application(s)
pharma/biopharma processes
storage temp.
15-25°C
SMILES string
O(C)c1cc(c(cc1)O)C(C)(C)C
InChI
1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3
InChI key
MRBKEAMVRSLQPH-UHFFFAOYSA-N
Application
Butylhydroxyanisole is an antioxidant and is typically used as an additive for the preservation of food.
Disclaimer
The substance 8.17023 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8170230000 | 04022536424866 |
| 8170231000 | 04022536424873 |
| 8170239025 | 04022536754529 |
