76233
Paraffin wax
tested according to Ph. Eur., low viscosity
Synonym(s):
Paraffinum Perliquidum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
application(s):
pharmaceutical (small molecule)
Recommended Products
Agency
USP/NF
tested according to Ph. Eur.
Quality Level
form
low viscosity
application(s)
pharmaceutical (small molecule)
InChI key
JWHAUXFOSRPERK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Immunohistochemical and Electron Microscopy Evolution in the Diagnosis of Lung Cancer in Iraq.: This study uses paraffin wax-embedded tissue samples for immunohistochemical and electron microscopy analyses to improve the diagnosis of lung cancer. Paraffin wax′s role in preserving tissue morphology and molecular integrity is crucial for accurate histological assessments (Saleem Abd Alkreem Alsaqi, 2022).
- Frequency of Expression of PD-1 and PD-L1 In Head And Neck Squamous Cell Carcinoma And their Association With Nodal Metastasis: A Cross-Sectional Study.: Utilizing paraffin wax-embedded samples, this study examines the expression of PD-1 and PD-L1 in head and neck squamous cell carcinoma, correlating their presence with nodal metastasis, thus aiding in the understanding of cancer progression and potential therapeutic targets (Malik et al., 2022).
- Evaluation of Cytopathological Techniques for the Diagnosis of Canine Visceral Leishmaniosis with Lymph Node Samples.: This research evaluates cytopathological techniques for diagnosing canine visceral leishmaniosis using paraffin wax-embedded lymph node samples, demonstrating the material′s importance in preserving tissue samples for accurate diagnostic analysis (Guerra et al., 2019).
- Systematic evaluation of PAXgene tissue fixation for the histopathological and molecular study of lung cancer.: The study compares PAXgene tissue fixation with traditional paraffin wax embedding for lung cancer tissues, highlighting paraffin wax′s reliability in maintaining tissue integrity for histopathological and molecular studies (Southwood et al., 2020).
- Semi-automated standardisation of melanin bleaching procedures of heavily pigmented melanocytic lesions for immunohistochemical analysis on an automated platform.: This study standardizes melanin bleaching procedures for heavily pigmented lesions using paraffin wax-embedded samples, facilitating accurate immunohistochemical analysis and improving diagnostic accuracy (Orchard et al., 2019).
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
602.6 °F - Pensky-Martens closed cup
Flash Point(C)
317 °C - Pensky-Martens closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Unni Cecilie Nygaard et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 123, 431-442 (2018-11-14)
Humans are daily exposed to mineral oil saturated hydrocarbons (MOSH) from the diet. We exposed female Fischer 344 rats to a broad mixture and sub-fractions of MOSH. Chemical characterization of the MOSH mixture used and material accumulated in rat tissues
Lindy Rauscher et al.
Journal of hazardous materials, 241-242, 441-449 (2012-10-24)
Surface waters impacted by urban runoff in metropolitan areas are becoming increasingly contaminated with polycyclic aromatic hydrocarbons (PAHs). Slow-release oxidant candles (paraffin-KMnO(4)) are a relatively new technology being used to treat contaminated groundwater and could potentially be used to treat
B Hilger et al.
Environmental pollution (Barking, Essex : 1987), 175, 16-21 (2013-01-15)
Levels and distribution of chlorinated paraffins were studied in randomly taken house dust samples (<63 μm) from the greater area of Munich, Germany. Quantification of short- (SCCPs, C(10)-C(13)) and medium-chain chlorinated paraffins (MCCPs, C(14)-C(16)) were performed using gas chromatography electron capture
Banu Dilek et al.
Archives of physical medicine and rehabilitation, 94(4), 642-649 (2012-11-29)
To evaluate the efficacy of paraffin bath therapy on pain, function, and muscle strength in patients with hand osteoarthritis. Prospective single-blinded randomized controlled trial. Department of physical medicine and rehabilitation in a university hospital. Patients with bilateral hand osteoarthritis (N=56).
Xue-Tong Wang et al.
Environmental science and pollution research international, 20(7), 4742-4749 (2013-01-08)
Short-chain chlorinated paraffins (SCCPs) are extremely complex technical mixtures of polychlorinated n-alkanes with carbon chain lengths from C10 to C13 and chlorine content between 49 and 70%. SCCPs are under consideration for inclusion in the Stockholm Convention on persistent organic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service