Skip to Content
Merck
All Photos(1)

Key Documents

411663

Sigma-Aldrich

Paraffin wax

mp ≥65 °C (ASTM D 87)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

beads or pellets
flakes or chunk(s)

Quality Level

mp

≥65 °C (ASTM D 87)

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Paraffin Wax is a low cost petroleum product that contains hydrocarbons with 20-40 carbon atoms. It is mainly used in lubricants, candles and electrical insulations.[1][2][3]

Application

Paraffin wax can be used as a phase changing material in a wide range of applications which include solar based water heaters,[4] microcapsules[5] and thermal energy devices (TEDs).[6]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paraffin Products, 14(, pages=) (1983)
Development of thermo-regulating textiles using paraffin wax microcapsules.
Sanchez P, et al.
Thermochimica Acta, 498(1-2), 16-21 (2010)
Performance investigation of thermal energy storage system with Phase Change Material (PCM) for solar water heating application.
Mahfuz MH, et al.
International Communications in Heat and Mass Transfer, 57(3), 132-139 (2014)
Heat transfer enhancement of paraffin wax using graphite foam for thermal energy storage.
Zhong Y, et al
Solar Energy Materials and Solar Cells, 94(6), 1011-1014 (2010)
The lubrication mechanism of calcium stearate/paraffin wax systems in PVC compounds.
Rabinovitch EB, et al.
Journal of Vinyl Technology, 6(3), 98-103 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service