Skip to Content
Merck
All Photos(3)

Key Documents

74731

Sigma-Aldrich

1-Octadecanethiol

purum, ≥95.0% (GC)

Synonym(s):

Mercaptan C18, Octadecyl mercaptan, Stearyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17SH
CAS Number:
Molecular Weight:
286.56
Beilstein:
1811934
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

purum

Quality Level

Assay

≥95.0% (GC)

form

solid

bp

204-210 °C/11 mmHg (lit.)

mp

30-33 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCS

InChI

1S/C18H38S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

QJAOYSPHSNGHNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octadecanethiol (ODT) is a water soluble sulfur compound with an 18 carbon alkyl chain, which forms a strong hydrophobic assembly with transition metals, like gold .It forms self-assembled monolayers (SAMs) on the surface of such transition metals. The thiol membrane when covalently bonded with the surface of the metal electrode, it shows electrostriction phenomenon leading to variation in capacitance.ODT SAMs have a hydrophobic surface with contact angle of 102oC.

Application

ODT is used to deposit thiolated molecules onto the surface of gold substrates, such as
  • SAMs on gold quartz crystal resonators.
  • on the tip of atomic force microscope probes.
  • capacitive sensors.

Analysis Note

may contain precipitation in melt

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polarization modulation Fourier transform infrared reflectance measurements of thin films and monolayers at metal surfaces utilizing real-time sampling electronics.
Barbara BJ, et al
Analytical Chemistry, 63(1), 55-60 (1991)
Jimin Yao et al.
Advanced materials (Deerfield Beach, Fla.), 22(10), 1102-1110 (2010-04-20)
Plasmonic crystals fabricated with precisely controlled arrays of subwavelength metal nanostructures provide a promising platform for sensing and imaging of surface binding events with micrometer spatial resolution over large areas. Soft nanoimprint lithography provides a robust, cost-effective method for producing
Ori Stein et al.
The journal of physical chemistry. B, 112(13), 3955-3962 (2008-03-08)
The interactions of water, carbon dioxide, and Xe with octadecanethiol (C(18)H(37)SH, ODT) self-assembled monolayers (SAMs) were studied under ultrahigh vacuum conditions employing temperature-programmed desorption and optical diffraction measurements. The ODT layer was grown on a 1 nm thick gold film
Khalid S Salaita et al.
Nano letters, 6(11), 2493-2498 (2006-11-09)
Experiments that utilize structures generated by dip-pen nanolithography (DPN) as positive resists for fabricating nanohole arrays and lithographic masters are described. The technique takes advantage of the difference in desorption potentials for patterned structures made from 16-mercaptohexadecanoic acid (MHA) and
Diego Millo et al.
Colloids and surfaces. B, Biointerfaces, 81(1), 212-216 (2010-08-03)
Hybrid bilayer lipid membranes (HBMs) were built on roughened silver electrodes exhibiting surface-enhanced Raman scattering (SERS) activity. The HBM consisted of a first layer of octadecanethiol (ODT) directly bound to the electrode surface, on which a second layer of 1,2-diphytanoyl-sn-glycero-3-phosphocholine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service