Skip to Content
Merck
All Photos(1)

Key Documents

70221

Supelco

Ammonium formate

eluent additive for LC-MS, LiChropur, ≥99.0%

Synonym(s):

Formic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2NH4
CAS Number:
Molecular Weight:
63.06
Beilstein:
3625095
EC Number:
MDL number:
UNSPSC Code:
12352107
eCl@ss:
39021903
PubChem Substance ID:
NACRES:
NB.21

grade

eluent additive for LC-MS

Quality Level

description

anionic

Assay

≥99.0% (calc. on dry substance, NT)
≥99.0%

form

crystals

quality

LiChropur

technique(s)

LC/MS: suitable

impurities

insoluble matter, passes filter test
<2% water

ign. residue (900 °C)

<0.1% (as SO4)

pH

5.5-7.5 (25 °C, 1 M in H2O)

mp

119-121 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.26 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤5 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤5 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.01
λ: 280 nm Amax: ≤0.01

SMILES string

N.OC=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium formate is the ammonium salt of formic acid. It is readily available, is stable and nontoxic in nature. It can be used in conjugation with Palladium on carbon (Pd-C) or Raney-Nickel catalysts.

Application

Ammonium formate has been used as buffer solution during LC-MS/MS analysis for screening of drugs and exogenous compounds. It may be used as a catalytic hydrogen transfer agent in reduction of aliphatic and aromatic nitro compounds.
Ammonium formate has been used in the separation of non-steroidal anti-inflammatory drugs and of oligosaccharide mixtures using capillary electrochromatography. It is also used in the analysis of phosphatidylserines, triacylglycerols and triacylglycerol oxidation products, and oligogalacturonic acids using HPLC and HPLC-MS methods.
Ammonium formate has been used as buffer solution during LC-MS/MS analysis for screening of drugs and exogenous compounds. It may be used as a catalytic hydrogen transfer agent in reduction of aliphatic and aromatic nitro compounds.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François-Ludovic Sauvage et al.
Clinical chemistry, 52(9), 1735-1742 (2006-07-22)
In clinical and forensic toxicology, general unknown screening is used to detect and identify exogenous compounds. In this study, we aimed to develop a comprehensive general unknown screening method based on liquid chromatography coupled with a hybrid triple-quadrupole linear ion
Lin Wang et al.
Annals of palliative medicine, 10(4), 4354-4368 (2021-05-11)
To explore the role of intestinal flora in seborrhea, non-targeted metabolomics analysis was carried out. Fecal samples were collected from 5 seborrheic patients and 5 healthy controls from October 2019 to April 2020. Ultra-high performance liquid chromatography-quadrupole time-of-flight mass spectrometry
A general procedure for mild and rapid reduction of aliphatic and aromatic nitro compounds using ammonium formate as a catalytic hydrogen transfer agent.
Ram, Siya, and Richard E. Ehrenkaufer.
Tetrahedron Letters, 25.32, 3415-3418 (1984)
Jiang-Tao Niu et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 4543761-4543761 (2020-12-22)
To compare the changes of chemical components of Hedysari Radix (HR) before and after honey-processing, and to explore the material basis of the difference between HR and honey-processed Hedysari Radix (HPHR) in Buzhong Yiqi. Different compounds in aqueous extracts of
Nara Liessi et al.
JCI insight, 5(16) (2020-07-17)
In recent years, a number of drugs have been approved for the treatment of cystic fibrosis (CF). Among them, newly released Trikafta, a combination of 3 drugs (VX-661/VX-445/VX-770), holds great promise to radically improve the quality of life for a

Articles

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

Ultra-high performance liquid chromatographic (UHPLC) separation of 12 nucleosides using Supel™ Carbon LC column in 15 minutes for use in identification and quantitation of nucleoside biomarkers.

See All

Chromatograms

application for HPLCapplication for HPLCapplication for HPLCapplication for HPLCShow More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service