Skip to Content
Merck
All Photos(1)

Key Documents

5.33001

Supelco

Acetic acid

100%, for LC-MS LiChropur

Synonym(s):

Acetic acid, Ethanoic acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
51242002
EC Index Number:
200-580-7
NACRES:
NB.21
Pricing and availability is not currently available.

Product Name

Acetic acid 100%, for LC-MS LiChropur

vapor density

2.07 (vs air)

Quality Level

vapor pressure

15.4 hPa ( 20 °C)

Assay

≥99.8% (acidimetric)

form

liquid

autoignition temp.

485 °C
800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

LC/MS: suitable

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L in H2O)

kinematic viscosity

1.17 cSt(20 °C)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

flash point 39 °C

density

1.04 g/mL at 25 °C (lit.)

cation traces

Al: ≤2.0 ppm
Ca: ≤20.0 ppm
Cu: ≤1.0 ppm
Fe: ≤20.0 ppm
K: ≤2.0 ppm
Mg: ≤2.0 ppm
NH4+: ≤10.0 ppm
Na: ≤5.0 ppm

storage temp.

15-25°C

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aceticacid 100% is the most commonly used additive or solvent in LC-MS (LiquidChromatography-Mass Spectrometry) applications.[1]

Application

Aceticacid 100% can be used as a background electrolyte (BGE) in CE-MS (capillaryelectrophoresis is coupled to mass spectrometry) analysis of peptide digests.[2]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klaus Faserl et al.
Methods (San Diego, Calif.), 184, 125-134 (2020-02-06)
The analysis, identification and quantification of histones and their post-translational modifications plays a central role in chromatin research and in studying epigenetic regulations during physiological processes. In the last decade analytical strategies based on mass spectrometry have been greatly improved
Risto Kostiainen et al.
Journal of chromatography. A, 1216(4), 685-699 (2008-09-19)
The most widely used ionization techniques in liquid chromatography-mass spectrometry (LC-MS) are electrospray ionization (ESI), atmospheric pressure chemical ionization (APCI) and atmospheric pressure photoionization (APPI). All three provide user friendly coupling of LC to MS. Achieving optimal LC-MS conditions is

Articles

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

An RP-LC×RP-LC-PDA-MS method for the metabolic profiling of G. glabra extract using the MSG approach for enhanced metabolic coverage.

See All

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service