Skip to Content
Merck
All Photos(1)

Documents

67126

Supelco

Oxolinic acid

analytical standard

Synonym(s):

5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO5
CAS Number:
Molecular Weight:
261.23
Beilstein:
620635
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

CCN1C=C(C(O)=O)C(=O)c2cc3OCOc3cc12

InChI

1S/C13H11NO5/c1-2-14-5-8(13(16)17)12(15)7-3-10-11(4-9(7)14)19-6-18-10/h3-5H,2,6H2,1H3,(H,16,17)

InChI key

KYGZCKSPAKDVKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Palominos et al.
Journal of hazardous materials, 158(2-3), 460-464 (2008-04-11)
This work studied the photocatalysed oxidation of the antibiotic oxolinic acid (OA) in an annular reactor operated with immobilized TiO(2) on sintered glass cylinders (SGC). Experiments were carried out in 1l solution of OA (18 mg l(-1)) at pH 9
gyrA and parC associated with quinolone resistance in Vibrio anguillarum.
C Rodkhum et al.
Journal of fish diseases, 31(5), 395-399 (2008-04-11)
Hong-Thih Lai et al.
Chemosphere, 75(4), 462-468 (2009-02-24)
Oxolinic acid (OA) and flumequine (FLU) are two of the quinolone antibiotics (QAs) that are widely used in aquaculture. The purpose of this study was to understand the fates of OA and FLU in waters and sediment slurries from aquaculture
Xiuhong Wang et al.
The Journal of antimicrobial chemotherapy, 65(3), 520-524 (2010-01-14)
Quinolone-mediated death of Escherichia coli has been proposed to occur by two pathways. One is blocked by inhibitors of protein synthesis; the other is not. It is currently unknown how these two pathways fit with the recent observation that hydroxyl
Alketa Tarushi et al.
Journal of inorganic biochemistry, 103(6), 898-905 (2009-04-28)
The neutral mononuclear zinc complexes with the quinolone antibacterial drug oxolinic acid in the absence or presence of a nitrogen donor heterocyclic ligand 2,2'-bipyridine or 1,10-phenanthroline have been synthesized and characterized. The experimental data suggest that oxolinic acid is on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service