Skip to Content
Merck
All Photos(1)

Documents

63043

Supelco

Magnesium matrix modifier

for graphite furnace-AAS

Synonym(s):

Magnesium nitrate – nitric acid solution, Magnesium modifier

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
4948473
MDL number:
UNSPSC Code:
12000000
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.21

grade

for graphite furnace-AAS

Quality Level

mol wt

Mr 256.41 g/mol

concentration

10.0 g/L±0.3 g/L Mg (in ~17% nitric acid)

cation traces

Ag: ≤0.001 mg/kg
As: ≤0.01 mg/kg
Cd: ≤0.001 mg/kg
Pb: ≤0.01 mg/kg
Se: ≤0.1 mg/kg
Tl: ≤0.01 mg/kg

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

General description

Matrix modification is a procedure for reducing volatilization and vapour-phase intrusions using a chemical reagent which acts as buffer enabling the use of higher pre-treated and atomization temperatures. The reagent causes either the interfering component to become more volatile or the analyte is converted to less volatile form. Magnesium is a first class modifier, forming oxides in the graphite furnace during heat pre-treatment. Magnesium added as nitrate has high thermal stability.

Application

Magnesium along with palladium may be used as matrix modifier during platform atomization, to determine lead in solution and in soil slurries, using electrothermal atomization atomic absorption spectrometry. It may also be used for determining brain manganese concentrations during graphite furnace atomic absorption spectrometry (GFAAS).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauri H. J. Lajunen, P. Peramaki
Spectrochemical Analysis by Atomic Absorption and Emission, 142-144 (2004)
Effectiveness of palladium plus magnesium as a matrix modifier for the determination of lead in solutions and soil slurries by electrothermal atomisation atomic absorption spectrometry.
Hinds, Michael W., Mohan Katyal, and Kenneth W. Jackson.
Journal of Analytical Atomic Spectrometry, 3, 83-87 (1988)
Magnesium nitrate as a matrix modifier in the stabilized temperature platform furnace
W. Slavin, G. R. Carnrick, and D. C. Manning
Analytical Chemistry, 54, 621-624 (1982)
Dinamene Santos et al.
Biological trace element research, 150(1-3), 337-341 (2012-09-14)
Manganese (Mn) is an essential element and it acts as a cofactor for a number of enzymatic reactions, including those involved in amino acid, lipid, protein, and carbohydrate metabolism. Excessive exposure to Mn can lead to poisoning, characterized by psychiatric
Zhichao Liu et al.
PLoS computational biology, 7(12), e1002310-e1002310 (2011-12-24)
Drug-induced liver injury (DILI) is a significant concern in drug development due to the poor concordance between preclinical and clinical findings of liver toxicity. We hypothesized that the DILI types (hepatotoxic side effects) seen in the clinic can be translated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service