Merck
Manganese alters rat brain amino acids levels.
Biological trace element research (2012-09-14)
Dinamene Santos, M Camila Batoreu, Isabel Almeida, Ruben Ramos, M Sidoryk-Wegrzynowicz, Michael Aschner, A P Marreilha dos Santos
ABSTRACT
Manganese (Mn) is an essential element and it acts as a cofactor for a number of enzymatic reactions, including those involved in amino acid, lipid, protein, and carbohydrate metabolism. Excessive exposure to Mn can lead to poisoning, characterized by psychiatric disturbances and an extrapyramidal disorder. Mn-induced neuronal degeneration is associated with alterations in amino acids metabolism. In the present study, we analyzed whole rat brain amino acid content subsequent to four or eight intraperitoneal injections, with 25 mg MnCl₂/kg/day, at 48-h intervals. We noted a significant increase in glycine brain levels after four or eight Mn injections (p < 0.05 and p < 0.01, respectively) and arginine also after four or eight injections (p < 0.001). Significant increases were also noted in brain proline (p < 0.01), cysteine (p < 0.05), phenylalanine (p < 0.01), and tyrosine (p < 0.01) levels after eight Mn injections vs. the control group. These findings suggest that Mn-induced alterations in amino acid levels secondary to Mn affect the neurochemical milieu.
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Supelco
Magnesium matrix modifier, for graphite furnace-AAS
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride solution, BioReagent, for molecular biology, storage temp.:room temp
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride 0.1 M solution