Skip to Content
Merck
All Photos(1)

Key Documents

57563

Supelco

Polydisperse Particle Standard

Particle Size Standard, PS224, 10 x 0.50g, 50-350 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

form

particles

manufacturer/tradename

Whitehouse Scientific Ltd PS224

particle size

50-350 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Polydisperse Particle Standard (PS224, size: 50-350 μm) is a particle size standard, ideal to profile particle size distribution (PSD) of test samples.
This product is characterized by techniques such as electroformed sieve analysis and microscopy.

Application

Used for particle size equipment calibration.
It can also be used to determine the precision and reproducibility of laser diffraction particle analyzer.

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • available as a neat sample
  • traceable to NIST and SRM standards

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guide to pharmaceutical product quality
Chemosphere, 53-83 (2018)
New insights on sources contributing dust to the loess record of the western edge of the Pampean Plain during the transition from the late MIS 2 to the early Holocene
Torre G, et al.
Chemosphere, 30(4), 537-545 (2020)
Revisiting the chronology and environmental conditions for the accretion of late Pleistocene-early Holocene Pampean loess (Argentina)
Torre G, et al.
Chemosphere, 213, 105-119 (2019)
The paleoclimatic message from the polymodal grain-size distribution of late Pleistocene-early Holocene Pampean loess (Argentina)
Torre G, et al.
Chemosphere, 42, 100563-100563 (2020)
Arsenic-bearing phases in South Andean volcanic ashes: Implications for As mobility in aquatic environments
Bia G, et al.
Chemical Geology , 393, 26-35 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service