Skip to Content
Merck
All Photos(1)

Key Documents

56753

Supelco

2-Phenoxyethanol

analytical standard

Synonym(s):

Ethylene glycol monophenyl ether, Phenylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1364011
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.539

bp

244-246 °C

mp

11-13 °C

density

1.107 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OCCOc1ccccc1

InChI

1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

QCDWFXQBSFUVSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenoxyethanol is a compound, which belongs to the class of volatile organicshaving a faint rose like aromatic odor. It may find applications as a solvent in ball-point inks, resins and in organic synthesis. It may also be used in cosmetics as an antimicrobial agent and preservative.
2-Phenoxyethanol is commonly used as an anesthetic in fisheries and aquaculture applications.

Application

2-Phenoxyethanol may be used as an analytical reference standard for the quantification of the analyte in pharmaceutical formulations and anesthetized fish using chromatography techniques. It may also be used as a test material to evaluate its efficacy as an anesthetic in juvenile European sea bass and gilthead sea bream through a series of experiments simulating aquaculture activities.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ash M
Handbook of Fillers, Extenders, and Diluents (2007)
A new validated HPLC method for the simultaneous determination of 2-phenoxyethanol, methylparaben, ethylparaben and propylparaben in a pharmaceutical gel
Shabir AG
Indian Journal of Pharmaceutical Sciences, 72(4), 421-421 (2010)
Development of an SPME?GC?MS/MS procedure for the monitoring of 2-phenoxyethanol in anaesthetised fish
Klimankova E, et al.
Talanta, 75(4), 1082-1088 (2008)
The identification of 2-phenoxyethanol in ballpoint inks using gas chromatography/mass spectrometry?relevance to ink dating
LaPorte, et al.
Journal of Forensic Sciences, 49(1), 1-5 (2003)
Comparative efficacy of clove oil and 2-phenoxyethanol as anesthetics in the aquaculture of European sea bass (Dicentrarchus labrax) and gilthead sea bream (Sparus aurata) at different temperatures
Mylonas CC, et al.
Aquaculture (Amsterdam, Netherlands), 246(1-4), 467-481 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service