Skip to Content
Merck
All Photos(1)

Key Documents

F0285200

Flurbiprofen

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flurbiprofen

manufacturer/tradename

EDQM

mp

110-112 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C(O)=O)c1ccc(c(F)c1)-c2ccccc2

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)

InChI key

SYTBZMRGLBWNTM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flurbiprofen EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor; non-steroidal anti-inflammatory agent with antifungal activity. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.
Fluibiprofen is a cyclooxygenase (COX) inhibitor, which is an enzyme responsible for the conversion of arachidonic acid to prostaglandin G2 (PGG2) and PGG2 to prostaglandin H2 (PGH2) in the prostaglandin synthesis pathway. This decreases the prostaglandins which cause inflammation, pain, swelling and fever. Flurbiprofen inhibits the activity of both COX-1 and -2. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Richy et al.
International journal of clinical practice, 61(8), 1396-1406 (2007-06-29)
The withdrawal of certain cyclooxygenase-2 selective drugs and the availability of over-the-counter non-steroidal anti-inflammatory drugs (NSAIDs) have increased the pressure for researching and prescribing conventional NSAIDs with a favourable efficacy/tolerance ratio in inflammatory diseases, particularly rheumatoid arthritis. The aim of
A Romano et al.
Journal of internal medicine, 241(1), 81-83 (1997-01-01)
Immune-mediated reactions to nonsteroidal anti-inflammatory drugs (NSAIDs) are unusual, and true allergy to the drug flurbiprofen has never been documented. We observed a patient who developed a maculopapular rash 48 h after beginning oral therapy with this drug, and 2
Asquad Sultan et al.
The Cochrane database of systematic reviews, (3)(3), CD007358-CD007358 (2009-07-10)
Flurbiprofen is a non-selective non-steroidal anti-inflammatory drug (NSAID), related to ibuprofen and naproxen, used to treat acute and chronic painful conditions. There is no systematic review of its use in acute postoperative pain. To assess efficacy, duration of action, and
Hugo Geerts
IDrugs : the investigational drugs journal, 10(2), 121-133 (2007-02-08)
(R)-flurbiprofen, the R-enantiomer of racemic flurbiprofen, is undergoing development by Myriad Genetics Inc, under license from Encore Pharmaceuticals Inc, for the potential treatment of Alzheimer's disease (AD). Devoid of any direct cyclooxygenase inhibition, which is associated with the more toxic
R N Brogden et al.
Drugs, 18(6), 417-438 (1979-12-01)
Flurbiprofen, a phenylalkanoic acid derivative, is a non-steroidal anti-inflammatory, antipyretic, analgesic agent advocated for use in rheumatoid arthritis, degenerative joint disease, ankylosing spondylitis and allied conditions. Published data suggest that flurbiprofen 120 to 150 mg daily is comparable in effectiveness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service