Skip to Content
Merck
All Photos(1)

Documents

53787

Supelco

Carazolol

analytical standard

Synonym(s):

1-(Carbazol-4-yloxy)-3-(isopropylamino)-2-propanol, 4-(2-Hydroxy-3-isopropylamino-propoxy)carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N2O2
CAS Number:
Molecular Weight:
298.38
Beilstein:
3620576
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

mp

133-137 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COc1cccc2[nH]c3ccccc3c12

InChI

1S/C18H22N2O2/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16/h3-9,12-13,19-21H,10-11H2,1-2H3

InChI key

BQXQGZPYHWWCEB-UHFFFAOYSA-N

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706030, T706025

Application

Carazolol may be used as a reference standard for the determination of the analyte:
  • In swine kidney by high-performance liquid chromatography with ultraviolet and fluorescence detection.
  • In animal tissues by high-performance liquid chromatography with electrochemical detection.

Commission Regulation (EU) No 37/2010 of 22 December 2009 on pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of tranquilisers and carazolol residues in animal tissue using high-performance liquid chromatography with electrochemical detection.
Rose MD and Shearer G
Journal of Chromatography A, 624(1-2), 471- 477 (1992)
Karin A Stephenson et al.
Journal of medicinal chemistry, 51(16), 5093-5100 (2008-07-29)
An efficient and general method has been developed for fluorine-18 labeling of beta-blockers that possess the propanolamine moiety. A new synthetically versatile intermediate, 3-(1-(benzyloxy)propan-2-yl)-2-oxooxazolidin-5-yl)methyl 4-methylbenzenesulfonate (13), was prepared and can be conjugated to any phenoxy core. To demonstrate the synthetic
Sid Topiol et al.
Biochemical pharmacology, 78(1), 11-20 (2009-05-19)
G-protein-coupled receptor (GPCR) proteins [Lundstrom KH, Chiu ML, editors. G protein-coupled receptors in drug discovery. CRC Press; 2006] are the single largest drug target, representing 25-50% of marketed drugs [Overington JP, Al-Lazikani B, Hopkins AL. How many drug targets are
Tomonaga Ozawa et al.
Bioorganic & medicinal chemistry, 19(17), 5231-5237 (2011-08-09)
We examined CH/π hydrogen bonds using an ab initio fragment molecular orbital (FMO) method, combined with the CHPI program, to evaluate complexes of active (bound with agonist 1) and inactive (bound with inverse agonist 2) β2 adrenergic receptor (β(2)AR) states.
Tony Warne et al.
Nature, 454(7203), 486-491 (2008-07-03)
G-protein-coupled receptors have a major role in transmembrane signalling in most eukaryotes and many are important drug targets. Here we report the 2.7 A resolution crystal structure of a beta(1)-adrenergic receptor in complex with the high-affinity antagonist cyanopindolol. The modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service