Skip to Content
Merck
All Photos(3)

Key Documents

52370

Sigma-Aldrich

Hexadecyltrimethylammonium bromide

≥96.0% (AT)

Synonym(s):

CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
Beilstein:
3598189
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

cationic

Quality Level

Assay

≥96.0% (AT)

form

powder or chunks

mol wt

364.45 g/mol

technique(s)

UV/Vis spectroscopy: suitable
electrophoresis: suitable

mp

248-251 °C (lit.)

solubility

H2O: 1 g/10 mL (warm)

application(s)

detection

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecyltrimethylammonium bromide is a titrant for the potentiometric titration of perchlorate with ionselective electrodes.

Application

<ul>
<li><strong>Sensitive detection in spectroscopy:</strong> The surface-active properties of Hexadecyltrimethylammonium bromide enhance the FRET effect in Cu-doped NH2-MIL-88(Fe), used for the highly sensitive detection of 3-nitro-L-tyrosine in biomolecular spectroscopy, contributing to advancements in diagnostic techniques and chemical analysis (Xu et al., 2024).</li>
<li><strong>Stabilization of heavy metals:</strong> Utilized in the intercalation-exfoliation modification of vermiculite for the stabilization of heavy metals during the pyrolysis of solid waste and sludge, Hexadecyltrimethylammonium bromide shows potential in waste management and pollution control, contributing to safer environmental practices (Yang et al., 2024).</li>
</ul>

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

471.2 °F - closed cup

Flash Point(C)

244 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Selig
Talanta, 26(11), 1061-1064 (1979-11-01)
Cetyltrimethylammonium bromide is used in the precipitation titration of perchlorate, with potentiometric end-point detection. A perchlorate, fluoroborate or nitrate ion-selective electrode may be used. The method is more sensitive than the commonly used titration with tetraphenylarsonium chloride. It can be
See Wee Chee et al.
Nature communications, 10(1), 2831-2831 (2019-06-30)
At elevated temperatures, bimetallic nanomaterials change their morphologies because of the interdiffusion of atomic species, which also alters their properties. The Kirkendall effect (KE) is a well-known phenomenon associated with such interdiffusion. Here, we show how KE can manifest in
Lourdes Pérez et al.
European journal of medicinal chemistry, 44(5), 1884-1892 (2008-12-17)
Biocompatible cationic surfactants from the amino acid lysine (hydrochloride salts of N(epsilon)-lauroyl lysine methyl ester, N(epsilon)-myristoyl lysine methyl ester and N(epsilon)-palmitoyl lysine methyl ester) have been prepared in high yields by lysine acylation in epsilon position with three natural saturated
Sebastien Fraud et al.
Antimicrobial agents and chemotherapy, 52(12), 4478-4482 (2008-10-08)
The biocide chlorhexidine (CHX) as well as additional membrane-active agents were shown to induce expression of the mexCD-oprJ multidrug efflux operon, dependent upon the AlgU stress response sigma factor. Hyperexpression of this efflux system in nfxB mutants was also substantially
Yu-Tsai Yang et al.
Lasers in surgery and medicine, 45(3), 175-185 (2013-03-20)
Antimicrobial photodynamic inactivation (PDI) is a promising therapeutic modality for the treatment of local infections. To increase the efficacy of PDI, chlorine e6 (Ce6) was encapsulated in cationic CTAB-liposomes composed of various ratios of dimyristoyl-sn-glycero-phosphatidylcholine (DMPC) and the cationic surfactant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service