Merck
  • Photodynamic inactivation of chlorin e6-loaded CTAB-liposomes against Candida albicans.
Photodynamic inactivation of chlorin e6-loaded CTAB-liposomes against Candida albicans.
Lasers in surgery and medicine (2013-03-20)
Yu-Tsai Yang, Hsiung-Fei Chien, Po-Han Chang, Yee-Chun Chen, Michael Jay, Tsuimin Tsai, Chin-Tin Chen
ABSTRACT
Antimicrobial photodynamic inactivation (PDI) is a promising therapeutic modality for the treatment of local infections. To increase the efficacy of PDI, chlorine e6 (Ce6) was encapsulated in cationic CTAB-liposomes composed of various ratios of dimyristoyl-sn-glycero-phosphatidylcholine (DMPC) and the cationic surfactant, cetyltrimethyl ammonium bromide (CTAB). The PDI efficacy of the liposomal-Ce6 was assessed in vitro against susceptible and drug-resistant clinical isolates of Candida albicans (C. albicans) as well as in infected burn wounds. Ce6 was encapsulated in CTAB-liposomes by the film hydration method. Particle size distribution and zeta potential of the cationic liposomes were measured using a Zetasizer Nano-ZS. UV-visible spectra were used to measure lipid/Ce6 (L/C) ratio and drug entrapment efficiency while differential scanning calorimetry (DSC) was used to study the thermotropic behavior of DMPC liposomes upon CTAB addition. In vivo PDI efficacy was carried out in an infected burn wound using a rat model. The increase in zeta potential and a shift in the phase transition temperature (Tm ) upon CTAB addition confirmed its entrapment within the lipid bilayers of the liposome. Meanwhile, the CTAB addition did not affect the Ce6 entrapment efficiency and physical attributes of the liposomes. In vitro studies showed that the PDI effect of the Ce6-loaded CTAB-liposomes was dependent on the lipid to Ce6 molar ratio (L/C), particle size and the concentration of CTAB in the liposomes. The lower L/C ratio and smaller liposomes exerted significantly higher PDI effects. In addition, an increase in the CTAB to lipid ratio led to a significant increase in the PDI effect of Ce6 against susceptible and drug-resistant clinical isolates of C. albicans after light illumination. Our results indicate that a low L/C ratio, high positive charge, and small particle size of CTAB-liposomes significantly enhances their PDI efficacy against C. albicans.
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)