Skip to Content
Merck
All Photos(2)

Key Documents

45657

Supelco

S 421

PESTANAL®, analytical standard

Synonym(s):

Bis(2,3,3,3-tetrachloropropyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl8O
CAS Number:
Molecular Weight:
377.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC(COCC(Cl)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

InChI

1S/C6H6Cl8O/c7-3(5(9,10)11)1-15-2-4(8)6(12,13)14/h3-4H,1-2H2

InChI key

LNJXZKBHJZAIKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S 421 belongs to the class of chloroalkyl ether compounds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
S 421 may be used as an analytical reference standard for the determination of S 421 in:
  • Tobacco samples by citrate QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction in acetonitrile solvent, low-temperature precipitation (LTP) and dispersive solid phase extraction (d-SPE) followed by analysis using gas chromatography combined with tandem mass spectrometry (GC-MS/MS) with electron impact (EI) ionization and selected reaction monitoring (SRM) detection.
  • Fresh tea shoots, tea infusion and spent tea leaves by GC coupled with 63Ni electron capture detector (ECD) and GC-MS operating on EI mode of detection.
  • Indoor air samples by GC-EI-MS equipped with selected ion monitoring (SIM) detection.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

350.6 °F

Flash Point(C)

177 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seisaku Yoshida et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 44(3), 175-179 (2003-09-13)
S-421 is a synergist for pyrethroid and organophosphorus pesticides, and is widely used in termiticides or commercial household insecticides. S-421 is mutagenic, stable and ubiquitous in the environment, and has been detected in household dust, surface water, sediments and rain-water.
Identification of the synergist bis(2,3,3,3-tetrachloropropyl)ether in human milk.
T Miyazaki et al.
Bulletin of environmental contamination and toxicology, 26(3), 420-423 (1981-03-01)
Robert I Krieger et al.
Environmental health perspectives, 111(12), 1439-1442 (2003-09-02)
Children and their parents in residences are often protected by insecticides from nuisance and disease-bearing mosquitoes. The annual worldwide consumption of the four major types of residential insecticide products--aerosols, mosquito coils, liquid vaporizers, and vaporizing mats--is in the billions of
Residues of the synergist S-421 in human milk collected from the Tokyo metropolitan area.
T Miyazaki
Bulletin of environmental contamination and toxicology, 29(5), 566-569 (1982-11-01)
An effective method for pesticide residues determination in tobacco by GC-MS/MS and UHPLC-MS/MS employing acetonitrile extraction with low-temperature precipitation and d-SPE clean-up.
Bernardi G, et al.
Talanta, 161(10), 40-47 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service