Skip to Content
Merck
All Photos(1)

Key Documents

34232

Supelco

Prothioconazole

PESTANAL®, analytical standard

Synonym(s):

2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15Cl2N3OS
CAS Number:
Molecular Weight:
344.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(CN1N=CNC1=S)(Cc2ccccc2Cl)C3(Cl)CC3

InChI

1S/C14H15Cl2N3OS/c15-11-4-2-1-3-10(11)7-14(20,13(16)5-6-13)8-19-12(21)17-9-18-19/h1-4,9,20H,5-8H2,(H,17,18,21)

InChI key

MNHVNIJQQRJYDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prothioconazole is a triazolinethione derivative, which can be used as a fungicide in order to inhibit the activity of demethylase enzyme.(1) It can be used in the treatment of infection in crops like wheat, caused by Mycosphaerella graminicola, a plant-pathogenic fungus.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel D A Tyndall et al.
PloS one, 11(12), e0167485-e0167485 (2016-12-03)
Azole antifungals, known as demethylase inhibitors (DMIs), target sterol 14α-demethylase (CYP51) in the ergosterol biosynthetic pathway of fungal pathogens of both plants and humans. DMIs remain the treatment of choice in crop protection against a wide range of fungal phytopathogens
Zhaoxian Zhang et al.
Journal of agricultural and food chemistry, 65(37), 8241-8247 (2017-08-29)
An efficient and sensitive chiral analytical method was established for the determination of the chiral fungicide prothioconazole and its major chiral metabolite prothioconazole-desthio in agricultural and environmental samples using ultraperformance liquid chromatography-tandem mass spectrometry. The optical rotation and absolute configuration
Carsten Albrecht Brühl et al.
Environmental science and pollution research international, 18(1), 31-37 (2010-06-15)
Seed treatments are widely used on cereals and other annual crops throughout Europe. Most of the formulated pesticide is found on the outside of the seed, the husk. Risk assessments of seed treatments are especially needed for granivorous mice living
Hyunkyu Sang et al.
Frontiers in microbiology, 9, 2335-2335 (2018-10-18)
The succinate dehydrogenase inhibitor (SDHI) fungicide, fluopyram, is used as a soybean seed treatment to manage Fusarium virguliforme, the casual agent of sudden death syndrome (SDS). More recently, other species within clade 2 of the Fusarium solani species, F. tucumaniae
Priscila Tessmer Scaglioni et al.
Journal of agricultural and food chemistry, 66(19), 4835-4841 (2018-04-28)
Fungicides and, for the first time, microalgal phenolic extracts (MPE) from Spirulina sp. and Nannochloropsis sp. were applied on maize culture media under field conditions to evaluate their ability to minimize Fusarium species development and fumonisin production. An in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service