Skip to Content
Merck
All Photos(1)

Key Documents

45456

Supelco

Diphenylamine

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
Beilstein:
508755
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.82 (vs air)

vapor pressure

1 mmHg ( 108 °C)

product line

PESTANAL®

autoignition temp.

1175 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

302 °C (lit.)

mp

50-53 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

N(c1ccccc1)c2ccccc2

InChI

1S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13H

InChI key

DMBHHRLKUKUOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenylamine is an organic compound, used for post-harvest deterioration of apple and pear crops. It finds applications in rubber production, dyes, polymers, pesticides, pharmaceutical, lubricating oils and photography chemicals.

Application

Diphenylamine may be used as an analytical reference standard for the quantification of the analyte in environmental samples and fruits using different analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

bottled under inert gas

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney,Liver,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative study between PCR and PLS in simultaneous spectrophotometric determination of diphenylamine, aniline, and phenol: effect of wavelength selection
Hemmateenejad B, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 67(2-4), 958-965 (2007)
Determination of ortho-phenylphenol, diphenyl and diphenylamine in apples and oranges using HPLC with fluorescence detection
Saad B, et al.
Food Chemistry, 84(2), 313-317 (2004)
M Francklin Philips et al.
Journal of hazardous materials, 237-238, 46-54 (2012-09-12)
Poly(diphenylamine-co-2-aminobenzonitrile) (P(DPA-co-2ABN)), a cyano group containing conducting polyaniline derivative, has been electrodeposited developed as the new material and utilized for the simultaneous electrochemical determination of trace levels of cadmium (Cd(2+)) and lead (Pb(2+)). P(DPA-co-2ABN) film preconcentrates effectively through cyano chelation
Katherine E Galvin-Burgess et al.
Stem cells (Dayton, Ohio), 31(1), 48-58 (2012-10-20)
Embryonic stem cells dynamically fluctuate between phenotypic states, as defined by expression levels of genes such as Nanog, while remaining pluripotent. The dynamic phenotype of stem cells is in part determined by gene expression control and dictated by various signaling
Tiffany Chang et al.
The Journal of clinical investigation, 123(1), 335-339 (2012-12-12)
Children with neurofibromatosis type 1 (NF1) are predisposed to juvenile myelomonocytic leukemia (JMML), an aggressive myeloproliferative neoplasm (MPN) that is refractory to conventional chemotherapy. Conditional inactivation of the Nf1 tumor suppressor in hematopoietic cells of mice causes a progressive MPN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service