Skip to Content
Merck
Diphenylamine PESTANAL®, analytical standard

Diphenylamine

Diphenylamine PESTANAL®, analytical standard
Synonym(s):
Diphenylamine
Linear Formula:
(C6H5)2NH
CAS Number:
Molecular Weight:
169.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique