Skip to Content
Merck
All Photos(3)

Key Documents

415049

Sigma-Aldrich

Aniline Blue diammonium salt

certified by the Biological Stain Commission

Synonym(s):

Ink Blue R; 4-[4-[[4-(4-aminooxysulfonylanilino)phenyl]-[4-(4-aminooxysulfonylphenyl)iminocyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H32N5O9S3
CAS Number:
Molecular Weight:
786.87
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder, crystals or granules

composition

Dye content, 65%

color

brown to very dark brown, and Red to Very Dark Red and Red-Brown and Very Dark Red-Brown

ε (extinction coefficient)

15000 at 595-605 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[S](=O)(=O)(ON)c1ccc(cc1)Nc2ccc(cc2)C(=C5C=CC(=Nc6ccc(cc6)[S](=O)(=O)ON)C=C5)c3ccc(cc3)Nc4ccc(cc4)[S](=O)(=O)O

InChI

1S/C37H31N5O9S3/c38-50-53(46,47)35-21-15-32(16-22-35)41-29-9-3-26(4-10-29)37(25-1-7-28(8-2-25)40-31-13-19-34(20-14-31)52(43,44)45)27-5-11-30(12-6-27)42-33-17-23-36(24-18-33)54(48,49)51-39/h1-24,40-41H,38-39H2,(H,43,44,45)

InChI key

CSYADFXDZYPANK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aniline Blue is the traditional name applied to anionic dyes produced by successive phenylation and sulfonation of basic fuchsine. It is a mixture of methyl blue (Cl 42780, Acid blue 93) and water blue (Cl 42755, Acid blue 22).

Application

Aniline blue (nominally as methyl blue or water blue) is a component of commonly used polychrome stains. Heidenhain′s AZAN stain and the Mallory′s polychrome stain both entail staining steps using aniline blue-orange G mixtures. Aniline blue is used in Gomori′s one-step trichrome stain and Mallory′s connective tissue stain for tissue including kidney and intestine.

Suitability

Certified for use in Mallory′s connective tissue stain and Gomori′s one-step trichrome.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Collagen XII Contributes to Epicardial and Connective Tissues in the Zebrafish Heart during Ontogenesis and Regeneration.
Marro J, et al.
PLoS ONE, 11(10) (2016)
Qin Peng et al.
International journal of molecular sciences, 21(4) (2020-02-16)
Oxathiapiprolin was developed as a specific plant pathogenic oomycete inhibitor, previously shown to have highly curative and protective activities against the pepper Phytophthora blight disease under field and greenhouse tests. Therefore, it was hypothesized that oxathiapiprolin might potentially activate the
Aniline blue and Calcofluor white staining of callose and cellulose in the streptophyte green algae Zygnema and Klebsormidium.
Herburger K and Holzinger A
Bio-protocol, 6(20) (2016)
Isolation and characterization of potential probiotic bacteria from pustulose ark (Anadara tuberculosa) suitable for shrimp farming
Sanchez-Ortiz A C, et al.
Latin American Journal of Aquatic Research, 43(1), 123-136 (2015)
Ya-Shiuan Lai et al.
Nature communications, 9(1), 3918-3918 (2018-09-27)
The unfolded protein response (UPR) of the endoplasmic reticulum constitutes a conserved and essential cytoprotective pathway designed to survive biotic and abiotic stresses that alter the proteostasis of the endoplasmic reticulum. The UPR is typically considered cell-autonomous and it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service