Skip to Content
Merck
All Photos(3)

Key Documents

HT15

Sigma-Aldrich

Trichrome Stain (Masson) Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

shelf life

Expiry date on the label

Quality Level

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Diagnostic Implications

May be useful in study of diseases of connective tissue and muscle characterized by fibrotic and dystrophic changes.

Principle

Collagen and muscle fiber stain differentially when treated sequentially with Biebrich Scarlet-Acid Fuchsin, PTA/PMA and Aniline Blue. Cytoplasm and muscle fibers stain red whereas collagen displays blue coloration.

Kit Components Only

Product No.
Description

  • Aniline Blue Solution (kit only) 250 mL

  • Biebrich Scarlet-Acid Fuchsin Solution (HT151) 250 mL

  • Phosphomolybdic Acid Solution (HT153) 250 mL

  • Phosphotungstic Acid Solution (HT152) 250 mL

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Ox. Liq. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O G Aztatzi-Aguilar et al.
Particle and fibre toxicology, 13(1), 68-68 (2016-12-14)
Particulate matter exposure is associated with respiratory and cardiovascular system dysfunction. Recently, we demonstrated that fine particles, also named PM2.5, modify the expression of some components of the angiotensin and bradykinin systems, which are involved in lung, cardiac and renal
Yu Liu et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 25(3), 666-678 (2017-02-02)
The promyelocytic leukemia protein (PML) is essential in the assembly of dynamic subnuclear structures called PML nuclear bodies (PML-NBs), which are involved in regulating diverse cellular functions. However, the possibility of PML being involved in cardiac disease has not been
Jiaxuan Chen et al.
Biomaterials, 113, 80-92 (2016-11-05)
Virtually all biomaterials are susceptible to biofilm formation and, as a consequence, device-associated infection. The concept of an immobilized liquid surface, termed slippery liquid-infused porous surfaces (SLIPS), represents a new framework for creating a stable, dynamic, omniphobic surface that displays
Meng-Huang Wu et al.
PloS one, 12(10), e0186784-e0186784 (2017-10-24)
This study evaluates the sustained analgesic effect of ketorolac-eluting thermosensitive biodegradable hydrogel in the plantar incisional pain model of the rat hind-paw. A ketorolac-embedded 2, 2'-Bis (2-oxazolin) (BOX) linking methoxy-poly(ethylene glycol) and poly(lactide-co-glycolide) (mPEG-PLGA) diblock copolymer (BOX copolymer) was synthesized
Clara Dees et al.
The Journal of experimental medicine, 208(5), 961-972 (2011-04-27)
Vascular damage and platelet activation are associated with tissue remodeling in diseases such as systemic sclerosis, but the molecular mechanisms underlying this association have not been identified. In this study, we show that serotonin (5-hydroxytryptamine [5-HT]) stored in platelets strongly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service