Skip to Content
Merck
All Photos(2)

Key Documents

39382

Supelco

Δ9-Tetrahydrocannabinolic acid A

analytical standard

Synonym(s):

THCA-A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (HPLC)

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule I; Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

suitability

corresponds for H-NMR

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CCCCCc1cc2OC(C)(C)C3CCC(C)=CC3c2c(O)c1C(O)=O

InChI

1S/C22H30O4/c1-5-6-7-8-14-12-17-19(20(23)18(14)21(24)25)15-11-13(2)9-10-16(15)22(3,4)26-17/h11-12,15-16,23H,5-10H2,1-4H3,(H,24,25)/t15-,16-/m1/s1

InChI key

UCONUSSAWGCZMV-HZPDHXFCSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Teixeira et al.
Forensic science international, 170(2-3), 148-155 (2007-07-07)
A fully validated, sensitive and specific method for the extraction and quantification of Delta(9)-tetrahydrocannabinol (THC) and 11-nor-9-carboxy-Delta(9)-THC (THC-COOH) and for the detection of 11-hydroxy-Delta(9)-THC (11-OH THC) in oral fluid, urine and whole blood is presented. Solid-phase extraction and liquid chromatography-mass
Rudolf Moldzio et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(8-9), 819-824 (2012-05-11)
Cannabinoids derived from Cannabis sativa demonstrate neuroprotective properties in various cellular and animal models. Mitochondrial impairment and consecutive oxidative stress appear to be major molecular mechanisms of neurodegeneration. Therefore we studied some major cannabinoids, i.e. delta-9-tetrahydrocannabinolic acid (THCA), delta-9-tetrahydrocannabinol (THC)
Marilyn A Huestis et al.
Clinical chemistry, 51(12), 2289-2295 (2005-10-15)
Knowing the time cannabis was last used is important for determining impairment in accident investigations and clinical evaluations. Two models for predicting time of last cannabis use from single plasma cannabinoid concentrations-model I, using Delta(9)-tetrahydrocannabinol (THC), and model II, using
Chandrasekaran Raman et al.
Amyotrophic lateral sclerosis and other motor neuron disorders : official publication of the World Federation of Neurology, Research Group on Motor Neuron Diseases, 5(1), 33-39 (2004-06-19)
Effective treatment for amyotrophic lateral sclerosis (ALS) remains elusive. Two of the primary hypotheses underlying motor neuron vulnerability are susceptibility to excitotoxicity and oxidative damage. There is rapidly emerging evidence that the cannabinoid receptor system has the potential to reduce
Garry Milman et al.
Journal of chromatography. A, 1217(9), 1513-1521 (2010-01-20)
Development and validation of a method for simultaneous identification and quantification of Delta9-tetrahydrocannabinol (THC), cannabidiol (CBD), cannabinol (CBN), and metabolites 11-hydroxy-THC (11-OH-THC) and 11-nor-9-carboxy-THC (THCCOOH) in oral fluid. Simultaneous analysis was problematic due to different physicochemical characteristics and concentration ranges.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service