Skip to Content
Merck
All Photos(2)

Key Documents

30958

Supelco

Benzo[b]fluoranthene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,3-Benzfluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, 3,4-Benzofluoranthene, Benz[e]acephenanthrylene, Benzo[e]fluoranthene, NSC 89265

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

163-165 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc-2c(c1)-c3cccc4c3c-2cc5ccccc45

InChI

1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H

InChI key

FTOVXSOBNPWTSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeh-Chung Chien et al.
Archives of toxicology, 84(10), 767-776 (2010-06-24)
Although urinary 1-hydroxypyrene (1-OHP) is the most relevant parameter for assessing exposure to polycyclic aromatic hydrocarbons, the inability to further elucidate the intra- and inter-individual variability, specificity and kinetics makes it difficult to enhance its value as an exposure predictor.
Hitoshi Ishikawa et al.
Cancer letters, 240(1), 29-35 (2005-10-26)
Air pollution has been suggested to cause genetic damage from investigations of many biological markers that measure cytogenetic damage in humans. Here, we evaluated the genotoxic effects of ambient air pollution by investigating the extent of cytogenetic damage in human
Vapor phase and particulate bound polycyclic aromatic hydrocarbons in the smoke of mosquito coils.
J M Lin et al.
Bulletin of environmental contamination and toxicology, 59(6), 868-874 (1997-12-24)
M J Mass et al.
Carcinogenesis, 17(8), 1701-1704 (1996-08-01)
The polycyclic aromatic hydrocarbon benzo[b]fluoranthene (B[b]F) is a pervasive constituent of environmental combustion products. We sought to examine the lung tumorigenic activity of B[b]F in strain A/J mice, to study the relationship between formation and decay of B[b]F-DNA adducts and
S Aubin et al.
Journal of the Air & Waste Management Association (1995), 50(12), 2093-2101 (2001-01-05)
Benzo[b]fluoranthene (B[b]F) was used in relative abundance ratios (RARs), a parameter obtained by dividing the concentration of individual polycyclic aromatic hydrocarbons (PAHs) found in a given sample by the concentration of B[b]F in the same sample. The B[b]F RARs were

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Compare PAH determination in olive oil: traditional GB 5009.265-2021 method vs. faster, simpler SPE with Supelclean EZ-POP NP; both meet criteria.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service