30498
N-Acetyl-S-phenyl-L-cysteine
analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.0% (HPLC)
optical activity
[α]/D -25.0±2.0°, c = 1 in ethanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
clinical testing
format
neat
SMILES string
CC(=O)N[C@@H](CSc1ccccc1)C(O)=O
InChI
1S/C11H13NO3S/c1-8(13)12-10(11(14)15)7-16-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3,(H,12,13)(H,14,15)/t10-/m0/s1
InChI key
CICOZWHZVMOPJS-JTQLQIEISA-N
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Recommended products
Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Wei sheng yan jiu = Journal of hygiene research, 39(5), 536-539 (2010-11-03)
To establish the method to determine S-phenylmercapturic acid (S-PMA) in human urine, and the method was actually applied to determine S-PMA concentrations in the human urine from subjects occupationally exposed to benzene. After the urine samples are hydrolyzed by acidification
Regulatory toxicology and pharmacology : RTP, 64(2 Suppl), S35-S44 (2012-09-04)
A randomized, controlled, open-label, parallel-group, single-center study to determine biomarkers of exposure to nine selected harmful and potentially harmful constituents (HPHC) in cigarette smoke and urinary excretion of mutagenic material in 160 male and female subjects smoking Marlboro cigarettes (6
The Annals of occupational hygiene, 55(3), 248-252 (2011-03-16)
This paper reports the assessment of worker exposure to high levels of benzene (up to 18 p.p.m. 8 h time weighted average, weekly average) during tunnelling on a contaminated land site (former gas works). Although respiratory and personal protection was
Toxicology letters, 213(1), 57-62 (2011-02-09)
The identification of reliable biomarkers is critical for the assessment of occupational exposure of benzene: S-phenylmercapturic acid (SPMA) and trans,trans-muconic acid (t,t-MA) are the most currently used. t,t-MA is an open-ring metabolite, but it is also a metabolite of the
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 16(4), 334-345 (2011-03-23)
Benzene is a ubiquitous pollutant; smoking habit, genetic polymorphisms, and analytical difficulties impact the identification of the best biomarker. To apply a systematic quantitative approach to evaluate urinary benzene (BEN-U) and S-phenylmercapturic acid (SPMA) as biomarkers of low benzene exposures.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service