Skip to Content
Merck
All Photos(4)

Key Documents

296309

Sigma-Aldrich

1,4-Dioxane

anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.8%
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

grade

anhydrous

Quality Level

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

99.8%

form

liquid

autoignition temp.

356 °F

expl. lim.

22 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

≤0.005%

refractive index

n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dioxane may undergo nickel-catalyzed oxidative arylation with arylboronic acids to form α-arylated ethers.
It may be used as a solvent to synthesize:
  • (Chloromethyl)dimethylphenylsilane from chloro(chloromethyl)dimethylsilane via zinc-catalyzed substitution reaction with phenylmagnesium bromide.
  • N-(2,2,2-trifluoro-1-hydroxyethyl)-acetamide from 2,2,2-trifluoro-1-methoxyethanol and acetamide.

Other Notes

The article number 296309-4X2L will be discontinued. Please order the single bottle 296309-2L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of tetraorganosilanes: (chloromethyl)dimethylphenylsilane
Murakami K, et al.
Organic Syntheses, 87, 178-178 (2010)
Solubility of anthracene in binary alcohol+ 1,4-dioxane solvent mixtures.
Powell JR, et al.
Journal of Chemical and Engineering Data, 40(5), 1124-1126 (1995)
A xanthate-transfer approach to ?-trifluoromethylamines
Gagosz F and Zard FS
Organic Syntheses, 84, 32-32 (2007)
Ignasi Casanellas et al.
Biomimetics (Basel, Switzerland), 4(2) (2019-06-28)
Extracellular matrix remodeling plays a pivotal role during mesenchyme patterning into different lineages. Tension exerted from cell membrane receptors bound to extracellular matrix ligands is transmitted by the cytoskeleton to the cell nucleus inducing gene expression. Here, we used dendrimer-based
Raquel Palao-Suay et al.
Macromolecular bioscience, 16(12), 1824-1837 (2016-10-16)
Well-structured amphiphilic copolymers are necessary to obtain self-assembled nanoparticles (NPs) based on synthetic polymers. Highly homogeneous and monodispersed macromolecules obtained by controlled polymerization have successfully been used for this purpose. However, disaggregation of the organized macromolecules is desired when a

Articles

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

Preformed catalysts in the kit are stable but become air-sensitive when activated; Schlenk technique aids scale-up.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service