Skip to Content
Merck
All Photos(3)

Key Documents

292249

Sigma-Aldrich

Boron trifluoride ethylamine complex

Synonym(s):

BF3-monoethylamine 1:1 complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · C2H5NH2
CAS Number:
Molecular Weight:
112.89
Beilstein:
3672802
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

mp

85-89 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN.FB(F)F

InChI

1S/C2H7N.BF3/c1-2-3;2-1(3)4/h2-3H2,1H3;

InChI key

JDVIRCVIXCMTPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boron trifluoride ethylamine complex may be used to initiate the curing of epoxy resins.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influences of inorganic fillers on curing reactions of epoxy resins initiated with a boron trifluoride amine complex.
Akatsuka M, et al.
Polymer, 42(7), 3003-3007 (2001)
Jean-Mathieu Pin et al.
ChemSusChem, 8(24), 4149-4161 (2015-12-15)
Epoxidized linseed oil and furfuryl alcohol are bio-sourced monomers known for their high-potential applications in materials science. In this work, we propose the association of these monomers through copolymerization reactions with the target to design fully bio-based thermosets. Herein, investigations
Synthesis and characterization of hybrid conducting composites based on polyaniline/magnetite fillers with improved microwave absorption properties
Belaabed B, et al.
J. Alloy Compounds, 527, 137-144 (2012)
Influences of inorganic fillers on curing reactions of epoxy resins initiated with a boron trifluoride amine complex
Akatsuka M, et al
Polymer, 42(7), 3003-3007 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service