Skip to Content
Merck
All Photos(1)

Documents

52649

Sigma-Aldrich

Hexamethylene diisocyanate

puriss., ≥99.0% (GC)

Synonym(s):

1,6-Diisocyanatohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCN(CH2)6NCO
CAS Number:
Molecular Weight:
168.19
Beilstein:
956709
EC Number:
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.25

grade

puriss.

Quality Level

Assay

≥99.0% (GC)

refractive index

n20/D 1.453

bp

82-85 °C/0.1 mmHg

density

1.047 g/mL at 20 °C (lit.)
1.047 g/mL at 20 °C

SMILES string

O=C=NCCCCCCN=C=O

InChI

1S/C8H12N2O2/c11-7-9-5-3-1-2-4-6-10-8-12/h1-6H2

InChI key

RRAMGCGOFNQTLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bifunctional reagent

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

266.0 °F - Pensky-Martens closed cup

Flash Point(C)

130 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Bieler et al.
Occupational medicine (Oxford, England), 61(6), 440-442 (2011-08-10)
Occupational diisocyanate-induced extrinsic allergic alveolitis (EAA) is a rare and probably underestimated diagnosis. Two acute occupational EAA cases have been described in this context, but neither of them concerned hexamethylene diisocyanate (HDI) exposure. To investigate the cause of a life-threatening
M Janko et al.
American Industrial Hygiene Association journal, 53(5), 331-338 (1992-05-01)
Monitoring of exposure to 1,6-hexamethylene (HDI) monomers and HDI polyisocyanates in Oregon was initiated in 1980 and covered primarily spray painting and related activities. A total of 562 air samples were collected from 60 workplaces during the years 1980-1990 and
Zibiao Li et al.
Biomacromolecules, 13(12), 3977-3989 (2012-11-22)
This paper reports the synthesis and characterization of new hyperbranched amphiphilic polyurethane multiblock copolymers consisting of poly(propylene glycol) (PPG), poly(ethylene glycol) (PEG), and polycaprolactone (PCL) segments as in situ thermogels. The hyperbranched poly(PPG/PEG/PCL urethane)s, termed as HBPEC copolymers, were synthesized
Sander M van Putten et al.
Journal of biomedical materials research. Part A, 98(4), 527-534 (2011-06-18)
Biomaterials are at continuous risk of bacterial contamination during production and application. In vivo, bacterial contamination of biomaterials delays the foreign body reaction (FBR). Endotoxins such as lipopolysaccharides (LPS), major constituents of the bacterial cell wall, are potent stimulators of
Victoria Arrandale et al.
International archives of occupational and environmental health, 86(2), 167-175 (2012-03-14)
Despite the importance of skin exposure, studies of skin symptoms in relation to exposure and respiratory symptoms are rare. The goals of this study were to describe exposure-response relationships for skin symptoms, and to investigate associations between skin and respiratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service