Skip to Content
Merck
All Photos(1)

Key Documents

25320

Sigma-Aldrich

1-Chloronaphthalene

technical, ≥85% (GC)

Synonym(s):

1-Naphthalenyl chloride, 1-Naphthyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Cl
CAS Number:
Molecular Weight:
162.62
Beilstein:
970836
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥85% (GC)

form

liquid

impurities

~10% 2-chloronaphthalene

refractive index

n20/D 1.632 (lit.)
n20/D 1.633

bp

111-113 °C/5 mmHg (lit.)

mp

−20 °C (lit.)

solubility

alcohol: soluble(lit.)
benzene-d6: soluble(lit.)
petroleum ether: soluble(lit.)

density

1.192 g/mL at 20 °C
1.194 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1cccc2ccccc12

InChI

1S/C10H7Cl/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

JTPNRXUCIXHOKM-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloronaphthalene (CN) improves the power conversion efficiency of bulk heterojunction solar cells. It forms 2:2 inclusion complex with β-cyclodextrin (β-CD), which is generated by the self-association of 1:1 β-CD-1CN inclusion complexes.

Application

1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was also used in preparation of PVC based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescence Polarization Studies of Inclusion Complexes between ?-Cyclodextrin and 1-Chloronaphthalene in Aqueous and Aqueous d-Glucose Solutions.
Hamai S.
The Journal of Physical Chemistry B, 103(2), 293-298 (1999)
Gisela L Schulz et al.
Beilstein journal of nanotechnology, 4, 680-689 (2013-11-10)
The optimization of solution-processed organic bulk-heterojunction solar cells with the acceptor-substituted quinquethiophene DCV5T-Bu 4 as donor in conjunction with PC61BM as acceptor is described. Power conversion efficiencies up to 3.0% and external quantum efficiencies up to 40% were obtained through
Ji Sun Moon et al.
Nano letters, 10(10), 4005-4008 (2010-09-10)
The bulk heterojunction (BHJ) material Si-PDTBT:PC(70)BM is sensitive to the use of a small amount of 1-chloronaphthalene (CN) as a processing additive; CN as a cosolvent (e.g., 4% in chlorobenzene) causes in a factor of 2 increase in the power
V K Gupta et al.
Talanta, 65(3), 730-734 (2008-10-31)
PVC based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) (TMOPP-Co) (I) as electroactive material with dibutyl butyl phosphonate (DBBP), dioctyl phthalate (DOP), 1-chloronaphthalene (CN), tri-n-butyl phosphate (TBP) and tris(2-ethylhexyl) phosphate (TEP) as plasticising solvent mediators have been prepared and tried for arsenite determination. The
[Isolated sensory trigeminal neuropathy. Discussion on possible toxic etiology (author's transl)].
P Gehin et al.
Revue d'oto-neuro-ophtalmologie, 53(3), 193-198 (1981-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service