Skip to Content
Merck
All Photos(2)

Key Documents

D116408

Sigma-Aldrich

2,7-Dihydroxynaphthalene

97%

Synonym(s):

2,7-Naphthalenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(OH)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
2042383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

185-190 °C (lit.)

SMILES string

Oc1ccc2ccc(O)cc2c1

InChI

1S/C10H8O2/c11-9-3-1-7-2-4-10(12)6-8(7)5-9/h1-6,11-12H

InChI key

DFQICHCWIIJABH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,7-Dihydroxynaphthalene is a organic building block used to prepare sulfonic acids, divinylnaphthalenes, dyes, pigments, and fluorescent whiteners.

Application

Starting material for the synthesis of sulfonic acids and divinylnaphthalenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raafat M Issa et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 980-986 (2005-06-14)
The absorption spectra of mono- and bis-azo-derivatives obtained by coupling the diazonium salts of aromatic amines and 2,7-dihydroxynaphthalene have been studied in six organic solvents. The different absorption bands have been assigned and the effect of solvents on the charge
C R Bloom et al.
Biochemistry, 36(42), 12746-12758 (1997-10-23)
The binding of phenolic ligands to the insulin hexamer occurs as a cooperative allosteric process. Investigations of the allosteric mechanism from this laboratory resulted in the postulation of a model consisting of a three-state conformational equilibrium and the derivation of
Subrata Kundu et al.
Langmuir : the ACS journal of surfaces and colloids, 26(9), 6720-6727 (2009-12-25)
A new synthetic route has been utilized for the formation of multiple-shaped Pt nanoparticles (NPs) under UV-photoirradiation. The one-step process exclusively generates different shapes, such as spheres, cubes, short and long wires, and flakelike nanostructures. The reduction of Pt(IV) ions
Maria Bonaccio et al.
Biochemistry, 44(21), 7656-7668 (2005-05-25)
The insulin hexamer is an allosteric protein widely used in formulations for the treatment of diabetes. The hexamer exhibits positive and negative cooperativity and apparent half-site binding activity, reflecting the interconversion of three allosteric states, designated as T6, T3R3, and
J. Chem. Soc. Perkin Trans. II, 721-721 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service