Skip to Content
Merck
All Photos(4)

Documents

179922

Sigma-Aldrich

Potassium phthalate monobasic

ACS reagent, acidimetric standard

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

99.95-100.05% dry basis

form

crystals

quality

acidimetric standard

impurities

≤0.002% S compounds
≤0.003% Cl compounds
≤0.005% insolubles

pH

4.00-4.02 (25.0 °C±0.2 °C, 0.05 M)

mp

295-300 °C (dec.) (lit.)

cation traces

Fe: ≤5 ppm
Na: ≤0.005%
heavy metals: ≤5 ppm (by ICP)

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Potassium phthalate monobasic may be used as a primary standard in acidimetric titration and also to prepare potassium phthalate buffer solution for pH calibration.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 4, 6-Trinitrobenzoic acid as a primary standerd in acidimetry.
Smith GF and Wilkins DH.
Analytica Chimica Acta, 8, 209-216 (1953)
Determination of standard pH values for potassium hydrogen phthalate reference buffer solutions in 10, 20, 50, 64 and 84.2 wt per cent methanol/water mixed solvents at temperatures from 283.15 to 313.15 K.
Mussini T, et al.
Electrochimica Acta, 28(11), 1593-1598 (1983)
J M Padró et al.
Talanta, 107, 95-102 (2013-04-20)
Dispersive ionic liquid-liquid microextraction combined with liquid chromatography and UV detection was used for the determination of two antichagasic drugs in human plasma: nifurtimox and benznidazole. The effects of experimental parameters on extraction efficiency-the type and volume of ionic liquid
Martha M Téllez-Rojo et al.
The Science of the total environment, 461-462, 386-390 (2013-06-12)
Previous studies suggest that prenatal phthalate exposure affects neurodevelopment and behavior during the first years of life. To evaluate the effect of maternal urinary concentrations of phthalate metabolites during pregnancy on mental and psychomotor development in children 24-36 months of
Randi J Bertelsen et al.
Environmental health perspectives, 121(2), 251-256 (2012-11-21)
High-molecular-weight phthalates in indoor dust have been associated with asthma in children, but few studies have evaluated phthalate biomarkers in association with respiratory outcomes. We explored the association between urinary concentrations of phthalate metabolites and current asthma. In a cross-sectional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service