Skip to Content
Merck
All Photos(1)

Key Documents

1.04874

Supelco

Potassium hydrogen phthalate

for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5KO4
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352106
EC Index Number:
212-889-4
NACRES:
NA.21
Assay:
≥99.5% dry basis (perchloric acid titration)
form:
solid
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

≥99.5% dry basis (perchloric acid titration)

form

solid

potency

>3200 mg/kg LD50, oral (Rat)

impurities

≤0.005% In water insoluble matter

loss

≤0.2% loss on drying, 105°C

pH

4.0 (20 °C, 50 g/L in H2O)

mp

295-300 °C

density

1.636 g/cm3

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfur compounds (as SO42-): ≤0.005%

cation traces

Cu: ≤0.0002%
Fe: ≤0.0005%
Na: ≤0.01%
Pb: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[O-]C(=O)c1c(cccc1)C(=O)[O-].[H+]

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Coulometric method with titratable impurity analysis and mass balance method: This study presents a method to convert acidimetric purity to chemical purity for potassium hydrogen phthalate, which is validated using quantitative nuclear magnetic resonance (qNMR) techniques, enhancing the standards for analytical chemistry (Huang T et al., 2023).

  • Triplet states of the nonlinear optical chromophore DCM in single crystals: Investigates the triplet excited states of the nonlinear optical chromophore DCM in potassium hydrogen phthalate single crystals, linking these states to single-molecule dark states, significant for the development of optical materials (Barbon A et al., 2009).


Analysis Note

Assay (Perchloric acid titration from dried substance): ≥ 99.5 %
Identity (IR-spectrum): passes test
In water insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.002 %
Sulfur compounds (as SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.0005 %
Loss on Drying (105°C): ≤ 0.2 %
Corresponds to Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service