Skip to Content
Merck
All Photos(1)

Documents

17773

Sigma-Aldrich

1-Amino-3-methylbutane hydrochloride

puriss., ≥98.0% (TLC)

Synonym(s):

Isopentylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
123.62
Beilstein:
3947083
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Quality Level

Assay

≥98.0% (TLC)

form

solid

mp

65-69 °C

SMILES string

Cl.CC(C)CCN

InChI

1S/C5H13N.ClH/c1-5(2)3-4-6;/h5H,3-4,6H2,1-2H3;1H

InChI key

HOMVDRDAAUYWKL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K S Ghenghesh et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 22(6), 653-665 (1989-01-01)
1. Gas liquid chromatography (GLC) was used to test the production of amines by 85 strains of Enterobacteriaceae. 2. The strains tested produced cadaverine, beta-phenylethylamine, putrescine, iso-amylamine, 2-methylbutylamine and iso-butylamine. 3. Although the overlap in amine production between obviously different
C Ji et al.
Carcinogenesis, 7(2), 301-303 (1986-02-01)
Nitrosomethylisoamylamine (NMIA), a carcinogenic N-nitroso compound was synthesized from isoamylamine (IAA) in a glucose-ammonium nitrate medium after several days' incubation with fungi and subsequent nitrosation with sodium nitrate. The nitrosamine was not produced by control reactions which lacked either IAA
[Studies on the synthesis of two new nitrosamines (NMAMBA and NMAMPA) from isoamylamine and sodium nitrite by fungi].
S J Lu et al.
Zhongguo yi xue ke xue yuan xue bao. Acta Academiae Medicinae Sinicae, 8(4), 255-260 (1986-08-01)
S Bu et al.
Biopharmaceutics & drug disposition, 21(4), 157-164 (2001-02-17)
In order to find what types of hepatic cytochrome P450 (CYP) isozymes are involved in the metabolism of 2-(allylthio)pyrazine (2-AP) in rats, enzyme inducers, such as phenobarbital, 3-methylcholanthrene, dexamethasone, or isoniazid, and an enzyme inhibitor, such as SKF 525-A were
Simon R Bailey et al.
American journal of veterinary research, 64(9), 1132-1138 (2003-09-19)
To measure concentrations of amines formed in the cecum of clinically normal ponies, determine amine concentrations in plasma samples collected in spring and winter, and compare concentrations of amines and serotonin in plasma samples obtained from clinically normal ponies and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service