Skip to Content
Merck
All Photos(1)

Key Documents

12479

Supelco

Silica gel Inorganic Sorbent

technical grade (w/ Ca, ~0.1%), 230-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

Product Name

Silica gel, technical grade (w/ Ca, ~0.1%), 60Å, 230-400 mesh particle size, Ca 0.1-0.3 %

grade

technical grade (w/ Ca, ~0.1%)

Quality Level

description

activity acc. to Brockmann and Schodder 2 - 3


for preparative liquid chromatography

form

powder

composition

Ca, 0.1-0.3%

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

230-400 mesh
40-63 μm

pore size

60 Å mean pore size

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel, highly porous textured matrix are generally used for synthesis of adsorbents. They are corpuscular in structure. Silica surface possess a layer of hydroxyl groups and adsorbed water molecules, which can be removed by drying at high temperature or even by treatment with suitable reagents. Silica gel surface plays important role in catalysis, chemical separation and microelectronic fabrications.

Application

  • Silica gel was used on glass plates for preparative TLC and on aluminium sheets for analytical TLC.
  • Silica gel was used in vacuum chromatography and TLC.
  • Silica gel was used in column chromatography.

Analysis Note

May contain insoluble matter and traces of process residues. Should be washed with solvent prior to use.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Jayaraman et al.
Chemosphere, 44(2), 181-191 (2001-07-11)
In this study, we compared microwave solvent extraction (MSE) to conventional methods for extracting organic contaminants from marine sediments and tissues with high and varying moisture content. The organic contaminants measured were polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, and polycyclic
Bioactive properties of wild blueberry fruits
Smith, M. A. L., et al
Journal of Food Science, 65, 352-356 (2000)
Proton NMR study of dehydration of the silica gel surface
Bronnimann
Journal of the American Chemical Society, 110, 2023-2026 (1988)
F. Daayf et al.
Plant physiology, 113(3), 719-727 (1997-03-01)
Phenolic compounds extracted from cucumber (Cucumis sativus L.) leaves were separated and analyzed for their differential presence and fungitoxicity in relation to a prophylactic treatment with Milsana (Compo, Munster, Germany) against powdery mildew (Sphaerotheca fuliginea). Based on our extraction and
The surface structure of silica gel
Peri, J. B., and A. L. Hensley Jr.
The Journal of Physical Chemistry, 72, 2926-2933 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service