Skip to Content
Merck

Skip To

89943

Silica gel Inorganic Sorbent

230-400 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Silica gel, suitable for column chromatography, 60

Quality Segment

form

powder

technique(s)

LPLC: suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

0.040-0.063 mm

pore size

60 Å mean pore size

bp

2230 °C

mp

>1600 °C

suitability

suitable for column chromatography

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

  • Flexible and Compressible Nanostructure-Assembled Aramid Nanofiber/Silica Composites Aerogel: This study explores the development of flexible and compressible silica-based aerogels, highlighting their potential in lightweight construction and thermal insulation applications, particularly useful for research in material science and industrial manufacturing (Zhang et al., 2024).
  • Factors Affecting Silica/Cellulose Nanocomposite Prepared via the Sol-Gel Technique: A Review: This review discusses the synthesis of silica/cellulose nanocomposites using the sol-gel method, emphasizing the impact of various factors on the material properties, relevant for researchers in materials science and environmental applications (Shange et al., 2024).
  • Spray-dried ternary bioactive glass microspheres: direct and indirect structural effects of copper-doping on acellular degradation behavior: Investigating the structural properties of copper-doped silica bioactive glass, this paper focuses on their applications in biomedicine and drug delivery systems, particularly in bone regeneration and tissue engineering (Vecchio et al., 2024).
  • Isolation, characterization, identification and quantification of 6-F oxyphenisatin dipropionate, a novel illegal additive, from a fruit-flavored jelly: Demonstrating the use of silica gel in chromatographic techniques for detecting illegal additives in food products, this study is vital for food safety and environmental testing laboratories (Xia et al., 2024).

Analysis Note

enriched with ~0.1% Ca to protect acid labile protection groups, actinty according to Brockmann and Schodder 2-3

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
607446074660745
matrix active group

silica

matrix active group

silica

matrix active group

silica

matrix active group

silica

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

separation technique

hydrophilic interaction (HILIC)

pore size

60 Å mean pore size

pore size

90 Å mean pore size

pore size

90 Å mean pore size

pore size

90 Å mean pore size

particle size

0.040-0.063 mm

particle size

0.015-0.025 mm, 15-25 μm

particle size

0.2-0.5 mm (35-70 mesh ASTM), 35-70 mesh

particle size

0.063-0.200 mm (70-230 mesh ASTM), 70-230 mesh

matrix

Silica

matrix

Silica

matrix

Silica

matrix

Silica

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable

technique(s)

LPLC: suitable


Still not finding the right product?

Explore all of our products under Silica gel


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters